Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92030281-92030532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX4
Entrez Gene ID
253152
Gene Name
epoxide hydrolase 4
Gene Aliases
ABHD7, EH4, EPHXRP
UniGene
Hs.201555
Gene Chromosome Location
Chr.1: 92029976-92063536 on Build GRCh38
Transcripts
NM_173567.4

SNP Detail

SNP ID Chromosome Location
rs749517168 Chr.1: 92030325-92030325 on Build GRCh38
rs868125445 Chr.1: 92030369-92030369 on Build GRCh38
rs564696748 Chr.1: 92030508-92030508 on Build GRCh38
rs780147383 Chr.1: 92030324-92030324 on Build GRCh38
rs1033120685 Chr.1: 92030395-92030395 on Build GRCh38
rs938453092 Chr.1: 92030477-92030477 on Build GRCh38
rs999332119 Chr.1: 92030381-92030381 on Build GRCh38
rs915571489 Chr.1: 92030480-92030487 on Build GRCh38
rs1056028060 Chr.1: 92030339-92030339 on Build GRCh38
rs542576596 Chr.1: 92030377-92030377 on Build GRCh38
rs777792858 Chr.1: 92030304-92030304 on Build GRCh38
rs5010338 Chr.1: 92030493-92030493 on Build GRCh38
rs957176703 Chr.1: 92030396-92030396 on Build GRCh38
rs55978071 Chr.1: 92030457-92030457 on Build GRCh38
rs5010335 Chr.1: 92030487-92030487 on Build GRCh38
rs574879324 Chr.1: 92030326-92030326 on Build GRCh38
rs183384568 Chr.1: 92030488-92030488 on Build GRCh38
rs371017117 Chr.1: 92030486-92030491 on Build GRCh38
rs910436788 Chr.1: 92030438-92030438 on Build GRCh38
rs985940895 Chr.1: 92030416-92030416 on Build GRCh38
rs991087000 Chr.1: 92030478-92030487 on Build GRCh38
rs374781868 Chr.1: 92030457-92030458 on Build GRCh38
rs371327373 Chr.1: 92030379-92030379 on Build GRCh38
rs1024722776 Chr.1: 92030371-92030371 on Build GRCh38
rs762970655 Chr.1: 92030397-92030397 on Build GRCh38
rs1014222975 Chr.1: 92030359-92030359 on Build GRCh38
rs563846633 Chr.1: 92030439-92030439 on Build GRCh38
rs745435867 Chr.1: 92030483-92030484 on Build GRCh38
rs755190160 Chr.1: 92030456-92030457 on Build GRCh38
rs773226063 Chr.1: 92030378-92030378 on Build GRCh38
rs867052011 Chr.1: 92030485-92030485 on Build GRCh38
rs756212462 Chr.1: 92030317-92030317 on Build GRCh38
rs928369696 Chr.1: 92030437-92030437 on Build GRCh38
rs894736481 Chr.1: 92030349-92030349 on Build GRCh38
rs5010337 Chr.1: 92030491-92030491 on Build GRCh38
rs918912190 Chr.1: 92030457-92030457 on Build GRCh38
rs768821963 Chr.1: 92030358-92030358 on Build GRCh38
rs185722847 Chr.1: 92030442-92030442 on Build GRCh38
rs752266917 Chr.1: 92030486-92030487 on Build GRCh38
rs113180520 Chr.1: 92030483-92030483 on Build GRCh38
rs5010336 Chr.1: 92030489-92030489 on Build GRCh38
rs746949723 Chr.1: 92030310-92030310 on Build GRCh38

View Full Product Details