Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193180608-193180853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT2
Entrez Gene ID
8707
Gene Name
beta-1,3-galactosyltransferase 2
Gene Aliases
BETA3GALT2, GLCT2, beta3Gal-T2
UniGene
Hs.518834
Gene Chromosome Location
Chr.1: 193178730-193186613 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4026323 c.895C>T p.P299S Chr.1: 193180668-193180668 on Build GRCh38
1184310 c.851G>A p.R284Q Chr.1: 193180712-193180712 on Build GRCh38
301736 c.919C>T p.R307C Chr.1: 193180644-193180644 on Build GRCh38
6229994 c.888G>A p.W296* Chr.1: 193180675-193180675 on Build GRCh38
4026324 c.866A>G p.N289S Chr.1: 193180697-193180697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371231651 Chr.1: 193180712-193180712 on Build GRCh38
rs753926771 Chr.1: 193180660-193180660 on Build GRCh38
rs374259217 Chr.1: 193180784-193180784 on Build GRCh38
rs572355338 Chr.1: 193180639-193180639 on Build GRCh38
rs770824791 Chr.1: 193180704-193180704 on Build GRCh38
rs146711525 Chr.1: 193180759-193180759 on Build GRCh38
rs767933476 Chr.1: 193180752-193180752 on Build GRCh38
rs377419023 Chr.1: 193180673-193180673 on Build GRCh38
rs761302481 Chr.1: 193180746-193180746 on Build GRCh38
rs748304059 Chr.1: 193180695-193180695 on Build GRCh38
rs780792859 Chr.1: 193180705-193180705 on Build GRCh38
rs774123687 Chr.1: 193180738-193180738 on Build GRCh38
rs745839994 Chr.1: 193180709-193180709 on Build GRCh38
rs762501860 Chr.1: 193180719-193180719 on Build GRCh38
rs956192261 Chr.1: 193180684-193180684 on Build GRCh38
rs1032346953 Chr.1: 193180652-193180652 on Build GRCh38
rs779324715 Chr.1: 193180687-193180687 on Build GRCh38
rs755649809 Chr.1: 193180772-193180772 on Build GRCh38
rs368427880 Chr.1: 193180829-193180829 on Build GRCh38
rs775377858 Chr.1: 193180713-193180713 on Build GRCh38
rs765402945 Chr.1: 193180817-193180817 on Build GRCh38
rs780301495 Chr.1: 193180649-193180649 on Build GRCh38
rs768145147 Chr.1: 193180732-193180732 on Build GRCh38
rs761273395 Chr.1: 193180763-193180763 on Build GRCh38
rs752978472 Chr.1: 193180825-193180825 on Build GRCh38

View Full Product Details