Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132572444-132572705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBRSL1
Entrez Gene ID
57666
Gene Name
fibrosin like 1
Gene Aliases
-
UniGene
Hs.411138
Gene Chromosome Location
Chr.12: 132490440-132585188 on Build GRCh38
Transcripts
NM_001142641.1 XM_005266171.3 XM_005266173.3 XM_005266175.3 XM_005266176.3 XM_005266177.3 XM_011534803.2 XM_011534804.2 XM_011534805.2 XM_011534806.2 XM_011534807.2 XM_011534808.2 XM_011534809.2 XM_011534810.2 XM_011534812.2 XM_011534816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298377 c.1584+9G>A p.? Chr.12: 132572631-132572631 on Build GRCh38
3954446 c.355-10T>C p.? Chr.12: 132572517-132572517 on Build GRCh38
6000501 c.1492T>C p.F498L Chr.12: 132572530-132572530 on Build GRCh38
6443454 c.1504C>T p.P502S Chr.12: 132572542-132572542 on Build GRCh38
937235 c.430C>T p.Q144* Chr.12: 132572602-132572602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769857540 Chr.12: 132572568-132572568 on Build GRCh38
rs769198659 Chr.12: 132572476-132572476 on Build GRCh38
rs760837740 Chr.12: 132572632-132572632 on Build GRCh38
rs779783064 Chr.12: 132572494-132572495 on Build GRCh38
rs191131759 Chr.12: 132572509-132572509 on Build GRCh38
rs909393498 Chr.12: 132572501-132572501 on Build GRCh38
rs775632498 Chr.12: 132572571-132572571 on Build GRCh38
rs904986562 Chr.12: 132572484-132572484 on Build GRCh38
rs1035504817 Chr.12: 132572673-132572675 on Build GRCh38
rs761406782 Chr.12: 132572634-132572634 on Build GRCh38
rs770762381 Chr.12: 132572654-132572654 on Build GRCh38
rs755081033 Chr.12: 132572508-132572508 on Build GRCh38
rs748369293 Chr.12: 132572534-132572534 on Build GRCh38
rs975329361 Chr.12: 132572665-132572665 on Build GRCh38
rs776411169 Chr.12: 132572514-132572514 on Build GRCh38
rs373932534 Chr.12: 132572500-132572500 on Build GRCh38
rs117416619 Chr.12: 132572582-132572582 on Build GRCh38
rs930549646 Chr.12: 132572646-132572646 on Build GRCh38
rs578132306 Chr.12: 132572482-132572482 on Build GRCh38
rs767876364 Chr.12: 132572648-132572648 on Build GRCh38
rs3829336 Chr.12: 132572499-132572499 on Build GRCh38
rs369215284 Chr.12: 132572567-132572567 on Build GRCh38
rs148231108 Chr.12: 132572470-132572470 on Build GRCh38
rs761107433 Chr.12: 132572658-132572658 on Build GRCh38
rs1044954591 Chr.12: 132572651-132572651 on Build GRCh38
rs940410746 Chr.12: 132572627-132572627 on Build GRCh38
rs375299122 Chr.12: 132572498-132572498 on Build GRCh38
rs920255815 Chr.12: 132572635-132572635 on Build GRCh38
rs746902434 Chr.12: 132572515-132572517 on Build GRCh38
rs750804583 Chr.12: 132572649-132572649 on Build GRCh38
rs964996527 Chr.12: 132572661-132572661 on Build GRCh38
rs1004043905 Chr.12: 132572493-132572493 on Build GRCh38
rs546036067 Chr.12: 132572628-132572628 on Build GRCh38
rs756640444 Chr.12: 132572496-132572496 on Build GRCh38
rs940833572 Chr.12: 132572529-132572529 on Build GRCh38
rs1050550830 Chr.12: 132572584-132572584 on Build GRCh38
rs377581541 Chr.12: 132572556-132572556 on Build GRCh38
rs201936807 Chr.12: 132572593-132572593 on Build GRCh38
rs770718344 Chr.12: 132572549-132572549 on Build GRCh38
rs866176175 Chr.12: 132572468-132572468 on Build GRCh38
rs866008592 Chr.12: 132572532-132572532 on Build GRCh38
rs774021267 Chr.12: 132572607-132572607 on Build GRCh38
rs992590627 Chr.12: 132572478-132572478 on Build GRCh38
rs998664535 Chr.12: 132572657-132572657 on Build GRCh38
rs987647453 Chr.12: 132572592-132572592 on Build GRCh38
rs1009545862 Chr.12: 132572503-132572503 on Build GRCh38

View Full Product Details