Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75675566-75675837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG4
Entrez Gene ID
1464
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
HMW-MAA, MCSP, MCSPG, MEL-CSPG, MSK16, NG2
UniGene
Hs.513044
Gene Chromosome Location
Chr.15: 75674322-75712848 on Build GRCh38
Transcripts
NM_001897.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6078015 c.6858C>A p.L2286L Chr.15: 75675661-75675661 on Build GRCh38
1608697 c.6827G>A p.R2276H Chr.15: 75675692-75675692 on Build GRCh38
6987904 c.6690C>A p.C2230* Chr.15: 75675829-75675829 on Build GRCh38
4556103 c.6799G>T p.G2267C Chr.15: 75675720-75675720 on Build GRCh38
1202511 c.6863C>G p.A2288G Chr.15: 75675656-75675656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779833639 Chr.15: 75675682-75675682 on Build GRCh38
rs530682211 Chr.15: 75675638-75675638 on Build GRCh38
rs768486425 Chr.15: 75675687-75675687 on Build GRCh38
rs934553944 Chr.15: 75675651-75675651 on Build GRCh38
rs773175028 Chr.15: 75675703-75675703 on Build GRCh38
rs201138980 Chr.15: 75675808-75675808 on Build GRCh38
rs766994775 Chr.15: 75675739-75675739 on Build GRCh38
rs747079269 Chr.15: 75675774-75675774 on Build GRCh38
rs3203767 Chr.15: 75675713-75675713 on Build GRCh38
rs771369858 Chr.15: 75675714-75675714 on Build GRCh38
rs766202843 Chr.15: 75675751-75675751 on Build GRCh38
rs749101671 Chr.15: 75675684-75675684 on Build GRCh38
rs761175264 Chr.15: 75675709-75675709 on Build GRCh38
rs186191601 Chr.15: 75675593-75675593 on Build GRCh38
rs772178990 Chr.15: 75675588-75675588 on Build GRCh38
rs756039904 Chr.15: 75675750-75675750 on Build GRCh38
rs563285050 Chr.15: 75675594-75675594 on Build GRCh38
rs777249049 Chr.15: 75675780-75675780 on Build GRCh38
rs988222528 Chr.15: 75675813-75675813 on Build GRCh38
rs545688154 Chr.15: 75675720-75675720 on Build GRCh38
rs148118120 Chr.15: 75675721-75675721 on Build GRCh38
rs748434385 Chr.15: 75675693-75675693 on Build GRCh38
rs995069406 Chr.15: 75675756-75675756 on Build GRCh38
rs1027580838 Chr.15: 75675809-75675809 on Build GRCh38
rs774145739 Chr.15: 75675726-75675726 on Build GRCh38
rs150766109 Chr.15: 75675773-75675773 on Build GRCh38
rs753811139 Chr.15: 75675757-75675757 on Build GRCh38
rs770273330 Chr.15: 75675793-75675793 on Build GRCh38
rs144719140 Chr.15: 75675652-75675652 on Build GRCh38
rs762651688 Chr.15: 75675648-75675648 on Build GRCh38
rs778736666 Chr.15: 75675764-75675764 on Build GRCh38
rs754763033 Chr.15: 75675763-75675763 on Build GRCh38
rs761463422 Chr.15: 75675814-75675814 on Build GRCh38
rs559710810 Chr.15: 75675665-75675665 on Build GRCh38
rs749908308 Chr.15: 75675742-75675742 on Build GRCh38
rs746448463 Chr.15: 75675789-75675789 on Build GRCh38
rs779075757 Chr.15: 75675692-75675692 on Build GRCh38
rs748307657 Chr.15: 75675766-75675766 on Build GRCh38
rs765597004 Chr.15: 75675725-75675725 on Build GRCh38
rs756800409 Chr.15: 75675655-75675655 on Build GRCh38
rs141405436 Chr.15: 75675770-75675770 on Build GRCh38
rs780637386 Chr.15: 75675664-75675664 on Build GRCh38
rs113030903 Chr.15: 75675737-75675737 on Build GRCh38
rs889823206 Chr.15: 75675733-75675733 on Build GRCh38
rs772175577 Chr.15: 75675702-75675702 on Build GRCh38
rs766556815 Chr.15: 75675804-75675804 on Build GRCh38
rs781124742 Chr.15: 75675741-75675741 on Build GRCh38
rs139353980 Chr.15: 75675779-75675779 on Build GRCh38
rs756019878 Chr.15: 75675673-75675673 on Build GRCh38

View Full Product Details