Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120129127-120129374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCN1
Entrez Gene ID
10985
Gene Name
GCN1, eIF2 alpha kinase activator homolog
Gene Aliases
GCN1L, GCN1L1, PRIC295
UniGene
Hs.298716
Gene Chromosome Location
Chr.12: 120127210-120194710 on Build GRCh38
Transcripts
NM_006836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4039353 c.7867C>T p.R2623W Chr.12: 120129299-120129299 on Build GRCh38
3456982 c.7829C>T p.A2610V Chr.12: 120129337-120129337 on Build GRCh38
7280041 c.7868G>A p.R2623Q Chr.12: 120129298-120129298 on Build GRCh38
935851 c.7808A>G p.D2603G Chr.12: 120129358-120129358 on Build GRCh38
6670050 c.7865T>C p.M2622T Chr.12: 120129301-120129301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771447541 Chr.12: 120129330-120129330 on Build GRCh38
rs548401692 Chr.12: 120129329-120129329 on Build GRCh38
rs368241252 Chr.12: 120129260-120129260 on Build GRCh38
rs961641275 Chr.12: 120129199-120129199 on Build GRCh38
rs978117209 Chr.12: 120129324-120129324 on Build GRCh38
rs777416354 Chr.12: 120129318-120129318 on Build GRCh38
rs775073318 Chr.12: 120129345-120129345 on Build GRCh38
rs756874630 Chr.12: 120129326-120129326 on Build GRCh38
rs766745731 Chr.12: 120129262-120129262 on Build GRCh38
rs770657729 Chr.12: 120129245-120129245 on Build GRCh38
rs1035147449 Chr.12: 120129259-120129260 on Build GRCh38
rs368130639 Chr.12: 120129348-120129348 on Build GRCh38
rs990871733 Chr.12: 120129294-120129294 on Build GRCh38
rs751663884 Chr.12: 120129293-120129293 on Build GRCh38
rs566164802 Chr.12: 120129156-120129156 on Build GRCh38
rs970967549 Chr.12: 120129234-120129234 on Build GRCh38
rs974266876 Chr.12: 120129263-120129263 on Build GRCh38
rs1005206686 Chr.12: 120129164-120129164 on Build GRCh38
rs749146416 Chr.12: 120129320-120129320 on Build GRCh38
rs765457878 Chr.12: 120129232-120129232 on Build GRCh38
rs1015713559 Chr.12: 120129173-120129173 on Build GRCh38
rs372483013 Chr.12: 120129300-120129300 on Build GRCh38
rs748858917 Chr.12: 120129194-120129194 on Build GRCh38
rs755055411 Chr.12: 120129299-120129299 on Build GRCh38
rs375253959 Chr.12: 120129313-120129313 on Build GRCh38
rs568435329 Chr.12: 120129260-120129260 on Build GRCh38
rs774159790 Chr.12: 120129347-120129347 on Build GRCh38
rs188100516 Chr.12: 120129328-120129328 on Build GRCh38
rs982368813 Chr.12: 120129202-120129202 on Build GRCh38
rs763457054 Chr.12: 120129349-120129349 on Build GRCh38
rs1002454239 Chr.12: 120129249-120129249 on Build GRCh38
rs1025678253 Chr.12: 120129208-120129208 on Build GRCh38
rs756090032 Chr.12: 120129315-120129315 on Build GRCh38

View Full Product Details