Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200593041-200593314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX1
Entrez Gene ID
316
Gene Name
aldehyde oxidase 1
Gene Aliases
AO, AOH1
UniGene
Hs.406238
Gene Chromosome Location
Chr.2: 200585952-200677064 on Build GRCh38
Transcripts
NM_001159.3 XM_011511062.1 XM_017003946.1 XM_017003947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6466816 c.61G>T p.V21F Chr.2: 200593161-200593161 on Build GRCh38
1641782 c.103+8T>C p.? Chr.2: 200593211-200593211 on Build GRCh38
6218853 c.85C>A p.P29T Chr.2: 200593185-200593185 on Build GRCh38
4847132 c.70G>A p.E24K Chr.2: 200593170-200593170 on Build GRCh38
5686630 c.98A>G p.K33R Chr.2: 200593198-200593198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755706342 Chr.2: 200593135-200593135 on Build GRCh38
rs779424881 Chr.2: 200593142-200593142 on Build GRCh38
rs764766851 Chr.2: 200593258-200593258 on Build GRCh38
rs746763518 Chr.2: 200593255-200593255 on Build GRCh38
rs758720037 Chr.2: 200593118-200593118 on Build GRCh38
rs766357746 Chr.2: 200593098-200593098 on Build GRCh38
rs961040118 Chr.2: 200593239-200593239 on Build GRCh38
rs752882911 Chr.2: 200593203-200593203 on Build GRCh38
rs1036466723 Chr.2: 200593082-200593082 on Build GRCh38
rs754130595 Chr.2: 200593100-200593100 on Build GRCh38
rs766277206 Chr.2: 200593174-200593174 on Build GRCh38
rs778379880 Chr.2: 200593125-200593125 on Build GRCh38
rs752175161 Chr.2: 200593208-200593208 on Build GRCh38
rs147623858 Chr.2: 200593104-200593104 on Build GRCh38
rs991019512 Chr.2: 200593078-200593078 on Build GRCh38
rs747447472 Chr.2: 200593127-200593127 on Build GRCh38
rs765121110 Chr.2: 200593117-200593120 on Build GRCh38
rs139174817 Chr.2: 200593204-200593204 on Build GRCh38
rs768435527 Chr.2: 200593148-200593148 on Build GRCh38
rs759566320 Chr.2: 200593188-200593188 on Build GRCh38
rs369319200 Chr.2: 200593109-200593109 on Build GRCh38
rs762779713 Chr.2: 200593222-200593225 on Build GRCh38
rs781740653 Chr.2: 200593210-200593210 on Build GRCh38
rs748827825 Chr.2: 200593219-200593219 on Build GRCh38
rs576029615 Chr.2: 200593164-200593164 on Build GRCh38
rs1045066997 Chr.2: 200593126-200593126 on Build GRCh38
rs774108736 Chr.2: 200593159-200593159 on Build GRCh38
rs894028050 Chr.2: 200593184-200593184 on Build GRCh38
rs940810837 Chr.2: 200593079-200593079 on Build GRCh38
rs142145534 Chr.2: 200593195-200593195 on Build GRCh38
rs771971157 Chr.2: 200593236-200593236 on Build GRCh38
rs747790866 Chr.2: 200593235-200593235 on Build GRCh38
rs778760266 Chr.2: 200593233-200593233 on Build GRCh38
rs760528585 Chr.2: 200593170-200593170 on Build GRCh38
rs747997929 Chr.2: 200593161-200593161 on Build GRCh38
rs1019265730 Chr.2: 200593231-200593231 on Build GRCh38
rs757709652 Chr.2: 200593209-200593209 on Build GRCh38
rs764349293 Chr.2: 200593207-200593207 on Build GRCh38
rs771741977 Chr.2: 200593163-200593163 on Build GRCh38
rs765388924 Chr.2: 200593105-200593105 on Build GRCh38
rs772889808 Chr.2: 200593250-200593250 on Build GRCh38
rs372717699 Chr.2: 200593146-200593146 on Build GRCh38
rs935599400 Chr.2: 200593176-200593176 on Build GRCh38
rs145888852 Chr.2: 200593220-200593220 on Build GRCh38
rs750286890 Chr.2: 200593214-200593214 on Build GRCh38
rs766363331 Chr.2: 200593226-200593227 on Build GRCh38
rs776707810 Chr.2: 200593179-200593179 on Build GRCh38
rs748737861 Chr.2: 200593145-200593145 on Build GRCh38

View Full Product Details