Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 48757883-48758154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTIF
Entrez Gene ID
9811
Gene Name
cap binding complex dependent translation initiation factor
Gene Aliases
Gm672, KIAA0427
UniGene
Hs.145230
Gene Chromosome Location
Chr.18: 48539056-48863217 on Build GRCh38
Transcripts
NM_001142397.1 NM_014772.2 XM_005258392.4 XM_006722583.3 XM_006722586.3 XM_006722587.3 XM_006722588.3 XM_006722589.3 XM_011526278.2 XM_011526279.2 XM_017026100.1 XM_017026101.1 XM_017026102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
75119 c.799G>A p.A267T Chr.18: 48758133-48758133 on Build GRCh38
4416436 c.637G>A p.D213N Chr.18: 48758115-48758115 on Build GRCh38
3526120 c.664A>G p.N222D Chr.18: 48758142-48758142 on Build GRCh38
6637448 c.795A>C p.A265A Chr.18: 48758129-48758129 on Build GRCh38
321174 c.458C>T p.P153L Chr.18: 48757936-48757936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371732858 Chr.18: 48757911-48757911 on Build GRCh38
rs755769161 Chr.18: 48757939-48757939 on Build GRCh38
rs754846409 Chr.18: 48758117-48758117 on Build GRCh38
rs776878929 Chr.18: 48758025-48758025 on Build GRCh38
rs755607388 Chr.18: 48758108-48758108 on Build GRCh38
rs751982639 Chr.18: 48757934-48757934 on Build GRCh38
rs899619124 Chr.18: 48758087-48758087 on Build GRCh38
rs148673663 Chr.18: 48758076-48758076 on Build GRCh38
rs549007210 Chr.18: 48758115-48758115 on Build GRCh38
rs775574826 Chr.18: 48758061-48758061 on Build GRCh38
rs1030905021 Chr.18: 48758085-48758085 on Build GRCh38
rs759634180 Chr.18: 48758035-48758035 on Build GRCh38
rs150490140 Chr.18: 48757904-48757904 on Build GRCh38
rs529167870 Chr.18: 48758079-48758079 on Build GRCh38
rs748009627 Chr.18: 48758020-48758020 on Build GRCh38
rs764733852 Chr.18: 48757929-48757929 on Build GRCh38
rs778949862 Chr.18: 48758001-48758001 on Build GRCh38
rs762340476 Chr.18: 48758077-48758077 on Build GRCh38
rs546795008 Chr.18: 48757962-48757962 on Build GRCh38
rs749164299 Chr.18: 48757960-48757960 on Build GRCh38
rs763215980 Chr.18: 48757920-48757920 on Build GRCh38
rs200095069 Chr.18: 48757970-48757970 on Build GRCh38
rs368257430 Chr.18: 48757906-48757906 on Build GRCh38
rs199863750 Chr.18: 48757910-48757910 on Build GRCh38
rs1033412889 Chr.18: 48758056-48758056 on Build GRCh38
rs989138919 Chr.18: 48758095-48758095 on Build GRCh38
rs756842456 Chr.18: 48757936-48757936 on Build GRCh38
rs770201937 Chr.18: 48758036-48758036 on Build GRCh38
rs943839062 Chr.18: 48758040-48758040 on Build GRCh38
rs764343360 Chr.18: 48758074-48758074 on Build GRCh38
rs757229684 Chr.18: 48758059-48758059 on Build GRCh38
rs533184947 Chr.18: 48757958-48757958 on Build GRCh38
rs763418953 Chr.18: 48758066-48758066 on Build GRCh38
rs142161540 Chr.18: 48758114-48758114 on Build GRCh38
rs202032754 Chr.18: 48758133-48758133 on Build GRCh38
rs374697225 Chr.18: 48757996-48757996 on Build GRCh38
rs772084041 Chr.18: 48758023-48758023 on Build GRCh38
rs111264927 Chr.18: 48757937-48757937 on Build GRCh38
rs942032486 Chr.18: 48758022-48758022 on Build GRCh38
rs923469324 Chr.18: 48757915-48757915 on Build GRCh38
rs957980839 Chr.18: 48758090-48758090 on Build GRCh38
rs999388776 Chr.18: 48758080-48758080 on Build GRCh38
rs372198425 Chr.18: 48758093-48758093 on Build GRCh38
rs749946917 Chr.18: 48758103-48758103 on Build GRCh38
rs151234284 Chr.18: 48758132-48758132 on Build GRCh38

View Full Product Details