Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200649081-200649306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX59
Entrez Gene ID
83479
Gene Name
DEAD-box helicase 59
Gene Aliases
OFD5, ZNHIT5
UniGene
Hs.497332
Gene Chromosome Location
Chr.1: 200640807-200669998 on Build GRCh38
Transcripts
NM_001031725.5 NM_001320181.1 NM_001320182.1 XM_005245519.1 XM_005245521.2 XM_006711562.1 XM_011510035.2 XM_017002431.1 XM_017002432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
901967 c.1349T>C p.F450S Chr.1: 200649192-200649192 on Build GRCh38
5969050 c.1334C>T p.P445L Chr.1: 200649207-200649207 on Build GRCh38
5969051 c.1334C>T p.P445L Chr.1: 200649207-200649207 on Build GRCh38
1668350 c.1334C>A p.P445H Chr.1: 200649207-200649207 on Build GRCh38
5969052 c.1334C>T p.P445L Chr.1: 200649207-200649207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576911947 Chr.1: 200649203-200649203 on Build GRCh38
rs748029526 Chr.1: 200649174-200649174 on Build GRCh38
rs761114437 Chr.1: 200649114-200649114 on Build GRCh38
rs147137498 Chr.1: 200649230-200649230 on Build GRCh38
rs143466956 Chr.1: 200649204-200649204 on Build GRCh38
rs751607825 Chr.1: 200649244-200649244 on Build GRCh38
rs6665604 Chr.1: 200649110-200649110 on Build GRCh38
rs754184314 Chr.1: 200649169-200649169 on Build GRCh38
rs201462184 Chr.1: 200649149-200649149 on Build GRCh38
rs756072457 Chr.1: 200649245-200649245 on Build GRCh38
rs138653543 Chr.1: 200649150-200649150 on Build GRCh38
rs147405057 Chr.1: 200649152-200649152 on Build GRCh38
rs368404651 Chr.1: 200649162-200649162 on Build GRCh38
rs147281351 Chr.1: 200649183-200649183 on Build GRCh38
rs115960805 Chr.1: 200649212-200649212 on Build GRCh38
rs79204709 Chr.1: 200649118-200649118 on Build GRCh38
rs536485578 Chr.1: 200649111-200649111 on Build GRCh38
rs755129562 Chr.1: 200649120-200649120 on Build GRCh38
rs771570708 Chr.1: 200649182-200649182 on Build GRCh38
rs912774739 Chr.1: 200649246-200649246 on Build GRCh38
rs759952696 Chr.1: 200649207-200649207 on Build GRCh38
rs374227657 Chr.1: 200649138-200649138 on Build GRCh38
rs769664474 Chr.1: 200649264-200649264 on Build GRCh38
rs749159795 Chr.1: 200649175-200649175 on Build GRCh38
rs761372457 Chr.1: 200649179-200649179 on Build GRCh38
rs754212550 Chr.1: 200649119-200649119 on Build GRCh38
rs963383109 Chr.1: 200649190-200649190 on Build GRCh38
rs768407092 Chr.1: 200649177-200649177 on Build GRCh38
rs150501573 Chr.1: 200649134-200649134 on Build GRCh38
rs758584419 Chr.1: 200649232-200649232 on Build GRCh38
rs17854157 Chr.1: 200649125-200649125 on Build GRCh38
rs1008342776 Chr.1: 200649217-200649217 on Build GRCh38
rs764189118 Chr.1: 200649236-200649236 on Build GRCh38
rs199598654 Chr.1: 200649151-200649151 on Build GRCh38
rs779911576 Chr.1: 200649249-200649249 on Build GRCh38
rs761185505 Chr.1: 200649187-200649187 on Build GRCh38
rs371608703 Chr.1: 200649178-200649178 on Build GRCh38

View Full Product Details