Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30497634-30497901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGCT
Entrez Gene ID
79017
Gene Name
gamma-glutamylcyclotransferase
Gene Aliases
C7orf24, CRF21, GCTG, GGC
UniGene
Hs.530024
Gene Chromosome Location
Chr.7: 30496621-30504841 on Build GRCh38
Transcripts
NM_001199815.1 NM_001199816.1 NM_001199817.1 NM_024051.3 NR_037669.1

SNP Detail

SNP ID Chromosome Location
rs201691947 Chr.7: 30497729-30497729 on Build GRCh38
rs920379283 Chr.7: 30497797-30497797 on Build GRCh38
rs539867008 Chr.7: 30497858-30497858 on Build GRCh38
rs745392114 Chr.7: 30497706-30497706 on Build GRCh38
rs778963174 Chr.7: 30497741-30497741 on Build GRCh38
rs554573697 Chr.7: 30497664-30497664 on Build GRCh38
rs1041115231 Chr.7: 30497757-30497757 on Build GRCh38
rs772323196 Chr.7: 30497737-30497737 on Build GRCh38
rs73083615 Chr.7: 30497661-30497661 on Build GRCh38
rs779240265 Chr.7: 30497720-30497720 on Build GRCh38
rs200273357 Chr.7: 30497739-30497739 on Build GRCh38
rs749488114 Chr.7: 30497724-30497724 on Build GRCh38
rs776495741 Chr.7: 30497764-30497764 on Build GRCh38
rs1031994569 Chr.7: 30497773-30497773 on Build GRCh38
rs1010633705 Chr.7: 30497683-30497683 on Build GRCh38
rs979861325 Chr.7: 30497704-30497704 on Build GRCh38
rs564723384 Chr.7: 30497794-30497794 on Build GRCh38
rs1006897543 Chr.7: 30497750-30497750 on Build GRCh38
rs999568265 Chr.7: 30497767-30497767 on Build GRCh38
rs956059426 Chr.7: 30497682-30497682 on Build GRCh38
rs200962598 Chr.7: 30497852-30497852 on Build GRCh38
rs901180848 Chr.7: 30497759-30497759 on Build GRCh38
rs762854515 Chr.7: 30497813-30497813 on Build GRCh38
rs539997363 Chr.7: 30497702-30497702 on Build GRCh38
rs755524545 Chr.7: 30497719-30497719 on Build GRCh38
rs10250220 Chr.7: 30497696-30497696 on Build GRCh38
rs775325678 Chr.7: 30497796-30497796 on Build GRCh38
rs763652057 Chr.7: 30497832-30497832 on Build GRCh38
rs889967712 Chr.7: 30497743-30497743 on Build GRCh38
rs540053508 Chr.7: 30497791-30497791 on Build GRCh38
rs746009360 Chr.7: 30497849-30497849 on Build GRCh38
rs1031628819 Chr.7: 30497671-30497671 on Build GRCh38
rs753074588 Chr.7: 30497659-30497659 on Build GRCh38
rs948427949 Chr.7: 30497698-30497698 on Build GRCh38
rs990028703 Chr.7: 30497695-30497695 on Build GRCh38

View Full Product Details