Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118901109-118901360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL9L
Entrez Gene ID
283149
Gene Name
B-cell CLL/lymphoma 9-like
Gene Aliases
BCL9-2, DLNB11
UniGene
Hs.414740
Gene Chromosome Location
Chr.11: 118896140-118926072 on Build GRCh38
Transcripts
NM_182557.2 XM_005271511.2 XM_005271512.2 XM_005271513.2 XM_006718815.2 XM_011542756.2 XM_017017572.1 XM_017017573.1 XM_017017574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6212949 c.2594A>G p.N865S Chr.11: 118901149-118901149 on Build GRCh38
428522 c.2385G>A p.L795L Chr.11: 118901358-118901358 on Build GRCh38
6894590 c.2455C>T p.R819W Chr.11: 118901288-118901288 on Build GRCh38
5458183 c.2438G>A p.S813N Chr.11: 118901305-118901305 on Build GRCh38
1351733 c.2432G>A p.G811D Chr.11: 118901311-118901311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779383973 Chr.11: 118901245-118901245 on Build GRCh38
rs776924640 Chr.11: 118901275-118901275 on Build GRCh38
rs749662780 Chr.11: 118901160-118901160 on Build GRCh38
rs780628293 Chr.11: 118901129-118901129 on Build GRCh38
rs748127006 Chr.11: 118901326-118901326 on Build GRCh38
rs778060405 Chr.11: 118901156-118901156 on Build GRCh38
rs143939490 Chr.11: 118901247-118901247 on Build GRCh38
rs748269018 Chr.11: 118901278-118901278 on Build GRCh38
rs781091194 Chr.11: 118901335-118901335 on Build GRCh38
rs531414289 Chr.11: 118901167-118901167 on Build GRCh38
rs866357789 Chr.11: 118901136-118901136 on Build GRCh38
rs780708177 Chr.11: 118901318-118901318 on Build GRCh38
rs768547300 Chr.11: 118901262-118901262 on Build GRCh38
rs762810833 Chr.11: 118901217-118901217 on Build GRCh38
rs148630206 Chr.11: 118901309-118901309 on Build GRCh38
rs374181286 Chr.11: 118901287-118901287 on Build GRCh38
rs780336341 Chr.11: 118901253-118901253 on Build GRCh38
rs753378628 Chr.11: 118901244-118901244 on Build GRCh38
rs368154927 Chr.11: 118901307-118901307 on Build GRCh38
rs772946389 Chr.11: 118901203-118901203 on Build GRCh38
rs771138812 Chr.11: 118901162-118901162 on Build GRCh38
rs183804575 Chr.11: 118901248-118901248 on Build GRCh38
rs755106983 Chr.11: 118901330-118901330 on Build GRCh38
rs746329697 Chr.11: 118901168-118901168 on Build GRCh38
rs547349167 Chr.11: 118901281-118901281 on Build GRCh38
rs1009995017 Chr.11: 118901181-118901181 on Build GRCh38
rs769979105 Chr.11: 118901338-118901338 on Build GRCh38
rs747103686 Chr.11: 118901261-118901261 on Build GRCh38
rs759038269 Chr.11: 118901317-118901317 on Build GRCh38
rs763468079 Chr.11: 118901288-118901288 on Build GRCh38
rs149942486 Chr.11: 118901226-118901226 on Build GRCh38
rs959876396 Chr.11: 118901267-118901267 on Build GRCh38
rs761025230 Chr.11: 118901184-118901184 on Build GRCh38
rs79075350 Chr.11: 118901174-118901174 on Build GRCh38
rs748441645 Chr.11: 118901337-118901337 on Build GRCh38
rs931320819 Chr.11: 118901319-118901319 on Build GRCh38
rs766026718 Chr.11: 118901219-118901219 on Build GRCh38
rs777487422 Chr.11: 118901240-118901240 on Build GRCh38
rs755771712 Chr.11: 118901154-118901154 on Build GRCh38
rs201347923 Chr.11: 118901341-118901341 on Build GRCh38
rs749949621 Chr.11: 118901272-118901272 on Build GRCh38
rs775340757 Chr.11: 118901282-118901282 on Build GRCh38
rs920622632 Chr.11: 118901147-118901147 on Build GRCh38
rs752375817 Chr.11: 118901138-118901138 on Build GRCh38
rs756831528 Chr.11: 118901294-118901294 on Build GRCh38
rs768908431 Chr.11: 118901196-118901196 on Build GRCh38
rs775683494 Chr.11: 118901182-118901182 on Build GRCh38
rs375686375 Chr.11: 118901291-118901291 on Build GRCh38
rs142121329 Chr.11: 118901314-118901314 on Build GRCh38
rs751516601 Chr.11: 118901224-118901224 on Build GRCh38
rs752192028 Chr.11: 118901231-118901231 on Build GRCh38
rs549521970 Chr.11: 118901232-118901232 on Build GRCh38
rs1032019058 Chr.11: 118901252-118901252 on Build GRCh38
rs770789853 Chr.11: 118901279-118901279 on Build GRCh38

View Full Product Details