Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116536108-116536339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD58
Entrez Gene ID
965
Gene Name
CD58 molecule
Gene Aliases
LFA-3, LFA3, ag3
UniGene
Hs.34341
Gene Chromosome Location
Chr.1: 116487140-116571093 on Build GRCh38
Transcripts
NM_001144822.1 NM_001779.2 NR_026665.1 XM_017002869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
220453 c.421C>T p.Q141* Chr.1: 116536172-116536172 on Build GRCh38
220553 c.392G>T p.C131F Chr.1: 116536201-116536201 on Build GRCh38
6623834 c.365-5delT p.? Chr.1: 116536233-116536233 on Build GRCh38
1200309 c.454C>T p.R152* Chr.1: 116536139-116536139 on Build GRCh38
5902445 c.485C>T p.P162L Chr.1: 116536108-116536108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151184981 Chr.1: 116536141-116536141 on Build GRCh38
rs199992898 Chr.1: 116536255-116536255 on Build GRCh38
rs183544787 Chr.1: 116536306-116536306 on Build GRCh38
rs565962590 Chr.1: 116536283-116536283 on Build GRCh38
rs750407102 Chr.1: 116536248-116536248 on Build GRCh38
rs41275570 Chr.1: 116536299-116536299 on Build GRCh38
rs1019945655 Chr.1: 116536222-116536222 on Build GRCh38
rs367618006 Chr.1: 116536218-116536218 on Build GRCh38
rs761758259 Chr.1: 116536232-116536232 on Build GRCh38
rs561957680 Chr.1: 116536233-116536233 on Build GRCh38
rs754559234 Chr.1: 116536138-116536138 on Build GRCh38
rs770324287 Chr.1: 116536200-116536200 on Build GRCh38
rs17850015 Chr.1: 116536154-116536154 on Build GRCh38
rs112685399 Chr.1: 116536301-116536301 on Build GRCh38
rs576382091 Chr.1: 116536297-116536297 on Build GRCh38
rs763294836 Chr.1: 116536214-116536214 on Build GRCh38
rs771503268 Chr.1: 116536165-116536165 on Build GRCh38
rs756010930 Chr.1: 116536249-116536249 on Build GRCh38
rs893436805 Chr.1: 116536199-116536199 on Build GRCh38
rs909869862 Chr.1: 116536307-116536307 on Build GRCh38
rs746356634 Chr.1: 116536189-116536189 on Build GRCh38
rs778504255 Chr.1: 116536146-116536146 on Build GRCh38
rs572700118 Chr.1: 116536233-116536233 on Build GRCh38
rs773787498 Chr.1: 116536232-116536233 on Build GRCh38
rs34546662 Chr.1: 116536149-116536149 on Build GRCh38
rs777232359 Chr.1: 116536188-116536188 on Build GRCh38
rs774492660 Chr.1: 116536220-116536220 on Build GRCh38
rs766224715 Chr.1: 116536254-116536254 on Build GRCh38
rs368947579 Chr.1: 116536265-116536265 on Build GRCh38
rs1056948524 Chr.1: 116536143-116536143 on Build GRCh38
rs200927101 Chr.1: 116536253-116536253 on Build GRCh38
rs951660836 Chr.1: 116536305-116536305 on Build GRCh38
rs999929633 Chr.1: 116536282-116536282 on Build GRCh38
rs1029883234 Chr.1: 116536300-116536300 on Build GRCh38
rs1018472579 Chr.1: 116536293-116536293 on Build GRCh38

View Full Product Details