Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 115178553-115178826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM1
Entrez Gene ID
23705
Gene Name
cell adhesion molecule 1
Gene Aliases
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
UniGene
Hs.370510
Gene Chromosome Location
Chr.11: 115173625-115504523 on Build GRCh38
Transcripts
NM_001098517.1 NM_001301043.1 NM_001301044.1 NM_001301045.1 NM_014333.3 XM_005271494.2 XM_017017457.1 XM_017017458.1 XM_017017459.1 XM_017017460.1 XM_017017461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5677048 c.1111G>A p.A371T Chr.11: 115178743-115178743 on Build GRCh38
686272 c.1085G>A p.R362Q Chr.11: 115178769-115178769 on Build GRCh38
4974253 c.1145C>T p.A382V Chr.11: 115178709-115178709 on Build GRCh38
6067654 c.1133G>T p.G378V Chr.11: 115178721-115178721 on Build GRCh38
2104081 c.1189C>T p.R397C Chr.11: 115178665-115178665 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765278524 Chr.11: 115178611-115178611 on Build GRCh38
rs761116141 Chr.11: 115178603-115178603 on Build GRCh38
rs148679370 Chr.11: 115178706-115178707 on Build GRCh38
rs774320439 Chr.11: 115178780-115178780 on Build GRCh38
rs138940776 Chr.11: 115178763-115178763 on Build GRCh38
rs758574980 Chr.11: 115178614-115178614 on Build GRCh38
rs757286282 Chr.11: 115178661-115178661 on Build GRCh38
rs775652567 Chr.11: 115178599-115178599 on Build GRCh38
rs745376752 Chr.11: 115178664-115178664 on Build GRCh38
rs752373195 Chr.11: 115178626-115178626 on Build GRCh38
rs1008446421 Chr.11: 115178590-115178590 on Build GRCh38
rs755207983 Chr.11: 115178703-115178703 on Build GRCh38
rs574717426 Chr.11: 115178613-115178613 on Build GRCh38
rs763412969 Chr.11: 115178766-115178766 on Build GRCh38
rs766626589 Chr.11: 115178621-115178621 on Build GRCh38
rs1012431388 Chr.11: 115178730-115178730 on Build GRCh38
rs755882347 Chr.11: 115178634-115178634 on Build GRCh38
rs777745290 Chr.11: 115178731-115178744 on Build GRCh38
rs765761542 Chr.11: 115178694-115178694 on Build GRCh38
rs543589576 Chr.11: 115178627-115178627 on Build GRCh38
rs773206839 Chr.11: 115178695-115178695 on Build GRCh38
rs199720777 Chr.11: 115178708-115178708 on Build GRCh38
rs11215399 Chr.11: 115178591-115178591 on Build GRCh38
rs762346837 Chr.11: 115178609-115178609 on Build GRCh38
rs746705418 Chr.11: 115178678-115178678 on Build GRCh38
rs200520615 Chr.11: 115178706-115178706 on Build GRCh38
rs992063283 Chr.11: 115178707-115178707 on Build GRCh38
rs767816262 Chr.11: 115178651-115178651 on Build GRCh38
rs777472577 Chr.11: 115178653-115178653 on Build GRCh38
rs952335280 Chr.11: 115178780-115178781 on Build GRCh38
rs61748616 Chr.11: 115178698-115178698 on Build GRCh38
rs867859915 Chr.11: 115178716-115178716 on Build GRCh38
rs766439518 Chr.11: 115178700-115178700 on Build GRCh38
rs145171991 Chr.11: 115178728-115178728 on Build GRCh38
rs771568327 Chr.11: 115178665-115178665 on Build GRCh38
rs779469616 Chr.11: 115178729-115178729 on Build GRCh38
rs763032683 Chr.11: 115178696-115178696 on Build GRCh38
rs757194461 Chr.11: 115178714-115178714 on Build GRCh38
rs771024754 Chr.11: 115178769-115178769 on Build GRCh38
rs767857191 Chr.11: 115178793-115178793 on Build GRCh38
rs35544534 Chr.11: 115178641-115178641 on Build GRCh38
rs759767344 Chr.11: 115178784-115178784 on Build GRCh38
rs141386367 Chr.11: 115178681-115178681 on Build GRCh38
rs764422872 Chr.11: 115178607-115178607 on Build GRCh38
rs267602705 Chr.11: 115178751-115178751 on Build GRCh38
rs552336980 Chr.11: 115178750-115178750 on Build GRCh38
rs745860759 Chr.11: 115178723-115178723 on Build GRCh38
rs775195967 Chr.11: 115178674-115178674 on Build GRCh38
rs146666877 Chr.11: 115178660-115178660 on Build GRCh38
rs374153442 Chr.11: 115178608-115178608 on Build GRCh38
rs777029275 Chr.11: 115178684-115178684 on Build GRCh38
rs200596202 Chr.11: 115178717-115178717 on Build GRCh38
rs772145243 Chr.11: 115178746-115178746 on Build GRCh38
rs762258727 Chr.11: 115178687-115178687 on Build GRCh38
rs773693771 Chr.11: 115178764-115178764 on Build GRCh38
rs748638917 Chr.11: 115178741-115178741 on Build GRCh38

View Full Product Details