Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87311103-87311344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf95
Entrez Gene ID
100506581
Gene Name
chromosome 16 open reading frame 95
Gene Aliases
-
UniGene
Hs.729380
Gene Chromosome Location
Chr.16: 87302798-87317420 on Build GRCh38
Transcripts
NM_001195124.1 NM_001195125.1 NM_001256917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6016333 c.352C>T p.P118S Chr.16: 87311275-87311275 on Build GRCh38
5422189 c.425A>T p.D142V Chr.16: 87311202-87311202 on Build GRCh38
6245488 c.391C>A p.H131N Chr.16: 87311236-87311236 on Build GRCh38
6342419 c.385C>A p.L129I Chr.16: 87311242-87311242 on Build GRCh38
6347165 c.444C>A p.Y148* Chr.16: 87311183-87311183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555008798 Chr.16: 87311186-87311186 on Build GRCh38
rs74038425 Chr.16: 87311261-87311261 on Build GRCh38
rs896233701 Chr.16: 87311293-87311293 on Build GRCh38
rs979335233 Chr.16: 87311252-87311252 on Build GRCh38
rs953488840 Chr.16: 87311213-87311213 on Build GRCh38
rs754130342 Chr.16: 87311286-87311286 on Build GRCh38
rs779349462 Chr.16: 87311199-87311199 on Build GRCh38
rs538323279 Chr.16: 87311311-87311311 on Build GRCh38
rs1014707885 Chr.16: 87311282-87311282 on Build GRCh38
rs867412641 Chr.16: 87311220-87311220 on Build GRCh38
rs947819536 Chr.16: 87311305-87311305 on Build GRCh38
rs777146365 Chr.16: 87311155-87311155 on Build GRCh38
rs561028660 Chr.16: 87311285-87311285 on Build GRCh38
rs1032851875 Chr.16: 87311165-87311165 on Build GRCh38
rs530478237 Chr.16: 87311237-87311237 on Build GRCh38
rs536353224 Chr.16: 87311184-87311184 on Build GRCh38
rs540816679 Chr.16: 87311214-87311214 on Build GRCh38
rs1056003379 Chr.16: 87311262-87311262 on Build GRCh38
rs756665178 Chr.16: 87311176-87311176 on Build GRCh38
rs148863765 Chr.16: 87311232-87311232 on Build GRCh38
rs933672743 Chr.16: 87311209-87311209 on Build GRCh38
rs759484340 Chr.16: 87311260-87311260 on Build GRCh38
rs903146641 Chr.16: 87311312-87311312 on Build GRCh38
rs752608423 Chr.16: 87311270-87311270 on Build GRCh38
rs181101593 Chr.16: 87311223-87311223 on Build GRCh38
rs562966151 Chr.16: 87311233-87311233 on Build GRCh38
rs963830652 Chr.16: 87311175-87311175 on Build GRCh38
rs752238043 Chr.16: 87311136-87311136 on Build GRCh38
rs558849785 Chr.16: 87311221-87311221 on Build GRCh38
rs776502999 Chr.16: 87311244-87311244 on Build GRCh38
rs140933864 Chr.16: 87311260-87311261 on Build GRCh38
rs899360354 Chr.16: 87311243-87311243 on Build GRCh38
rs1038489379 Chr.16: 87311242-87311242 on Build GRCh38
rs769089257 Chr.16: 87311193-87311193 on Build GRCh38
rs758078221 Chr.16: 87311137-87311137 on Build GRCh38
rs368517062 Chr.16: 87311321-87311321 on Build GRCh38
rs145644540 Chr.16: 87311206-87311206 on Build GRCh38
rs887401046 Chr.16: 87311318-87311318 on Build GRCh38
rs751141070 Chr.16: 87311164-87311164 on Build GRCh38
rs967484136 Chr.16: 87311131-87311131 on Build GRCh38

View Full Product Details