Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151646523-151646796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160A1
Entrez Gene ID
729830
Gene Name
family with sequence similarity 160 member A1
Gene Aliases
-
UniGene
Hs.633810
Gene Chromosome Location
Chr.4: 151409143-151670503 on Build GRCh38
Transcripts
NM_001109977.1 XM_005263199.1 XM_011532220.1 XM_011532221.2 XM_011532222.2 XM_011532223.2 XM_011532224.2 XM_017008583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594360 c.1372G>T p.E458* Chr.4: 151646703-151646703 on Build GRCh38
5400623 c.1377G>A p.R459R Chr.4: 151646708-151646708 on Build GRCh38
7333914 c.1227G>T p.R409S Chr.4: 151646558-151646558 on Build GRCh38
5474676 c.1330G>A p.V444I Chr.4: 151646661-151646661 on Build GRCh38
5034082 c.1405C>A p.H469N Chr.4: 151646736-151646736 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192710881 Chr.4: 151646741-151646741 on Build GRCh38
rs370066018 Chr.4: 151646690-151646690 on Build GRCh38
rs867003111 Chr.4: 151646572-151646572 on Build GRCh38
rs576546743 Chr.4: 151646554-151646554 on Build GRCh38
rs955505269 Chr.4: 151646551-151646551 on Build GRCh38
rs762798286 Chr.4: 151646619-151646620 on Build GRCh38
rs759129776 Chr.4: 151646579-151646579 on Build GRCh38
rs750468420 Chr.4: 151646675-151646675 on Build GRCh38
rs529874295 Chr.4: 151646729-151646729 on Build GRCh38
rs188094362 Chr.4: 151646574-151646574 on Build GRCh38
rs773850578 Chr.4: 151646576-151646576 on Build GRCh38
rs370765283 Chr.4: 151646615-151646615 on Build GRCh38
rs775802962 Chr.4: 151646642-151646642 on Build GRCh38
rs372755280 Chr.4: 151646676-151646676 on Build GRCh38
rs1004404642 Chr.4: 151646759-151646759 on Build GRCh38
rs77353765 Chr.4: 151646770-151646770 on Build GRCh38
rs890520353 Chr.4: 151646678-151646678 on Build GRCh38
rs16998842 Chr.4: 151646758-151646758 on Build GRCh38
rs917656035 Chr.4: 151646561-151646561 on Build GRCh38
rs765246520 Chr.4: 151646672-151646672 on Build GRCh38
rs899829050 Chr.4: 151646710-151646710 on Build GRCh38
rs770341175 Chr.4: 151646635-151646635 on Build GRCh38
rs143316272 Chr.4: 151646636-151646636 on Build GRCh38
rs374252624 Chr.4: 151646599-151646599 on Build GRCh38
rs775872315 Chr.4: 151646603-151646603 on Build GRCh38
rs552938541 Chr.4: 151646736-151646736 on Build GRCh38
rs1016906997 Chr.4: 151646570-151646570 on Build GRCh38
rs970466950 Chr.4: 151646562-151646562 on Build GRCh38
rs762317274 Chr.4: 151646658-151646658 on Build GRCh38
rs929276361 Chr.4: 151646648-151646648 on Build GRCh38
rs752591226 Chr.4: 151646731-151646731 on Build GRCh38
rs1039167349 Chr.4: 151646689-151646689 on Build GRCh38
rs377280064 Chr.4: 151646560-151646560 on Build GRCh38
rs937294108 Chr.4: 151646628-151646628 on Build GRCh38
rs764642044 Chr.4: 151646649-151646649 on Build GRCh38
rs941906176 Chr.4: 151646581-151646581 on Build GRCh38
rs377024303 Chr.4: 151646683-151646683 on Build GRCh38

View Full Product Details