Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150751554-150751825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSS
Entrez Gene ID
1520
Gene Name
cathepsin S
Gene Aliases
-
UniGene
Hs.181301
Gene Chromosome Location
Chr.1: 150730196-150765957 on Build GRCh38
Transcripts
NM_001199739.1 NM_004079.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5353681 c.603C>T p.D201D Chr.1: 150751805-150751805 on Build GRCh38
462914 c.601G>C p.D201H Chr.1: 150751807-150751807 on Build GRCh38
3473487 c.615C>T p.P205P Chr.1: 150751793-150751793 on Build GRCh38
4675487 c.595G>A p.D199N Chr.1: 150751813-150751813 on Build GRCh38
4771015 c.624C>G p.A208A Chr.1: 150751784-150751784 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770467962 Chr.1: 150751777-150751777 on Build GRCh38
rs376976448 Chr.1: 150751804-150751804 on Build GRCh38
rs115824602 Chr.1: 150751805-150751805 on Build GRCh38
rs773881952 Chr.1: 150751790-150751790 on Build GRCh38
rs1016934855 Chr.1: 150751600-150751600 on Build GRCh38
rs969756858 Chr.1: 150751634-150751634 on Build GRCh38
rs927842573 Chr.1: 150751768-150751768 on Build GRCh38
rs1026334135 Chr.1: 150751701-150751701 on Build GRCh38
rs140288328 Chr.1: 150751665-150751665 on Build GRCh38
rs112975544 Chr.1: 150751694-150751694 on Build GRCh38
rs1016560411 Chr.1: 150751758-150751758 on Build GRCh38
rs372315946 Chr.1: 150751763-150751763 on Build GRCh38
rs950654364 Chr.1: 150751712-150751712 on Build GRCh38
rs16840587 Chr.1: 150751772-150751772 on Build GRCh38
rs1002079166 Chr.1: 150751585-150751585 on Build GRCh38
rs587715551 Chr.1: 150751776-150751776 on Build GRCh38
rs896596641 Chr.1: 150751661-150751661 on Build GRCh38
rs192282333 Chr.1: 150751700-150751700 on Build GRCh38
rs780363637 Chr.1: 150751740-150751740 on Build GRCh38
rs1006486355 Chr.1: 150751736-150751736 on Build GRCh38

View Full Product Details