Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55352061-55352324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO34
Entrez Gene ID
55030
Gene Name
F-box protein 34
Gene Aliases
CGI-301, Fbx34
UniGene
Hs.525348
Gene Chromosome Location
Chr.14: 55271303-55355222 on Build GRCh38
Transcripts
NM_017943.3 NM_152231.1 XM_005267791.4 XM_006720185.3 XM_017021391.1 XM_017021392.1 XM_017021393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6140864 c.1801A>G p.T601A Chr.14: 55352191-55352191 on Build GRCh38
136973 c.1883G>A p.R628K Chr.14: 55352273-55352273 on Build GRCh38
7212726 c.1782T>G p.S594R Chr.14: 55352172-55352172 on Build GRCh38
6045998 c.1840A>G p.I614V Chr.14: 55352230-55352230 on Build GRCh38
6140865 c.1702C>G p.L568V Chr.14: 55352092-55352092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61755586 Chr.14: 55352295-55352295 on Build GRCh38
rs984794818 Chr.14: 55352174-55352174 on Build GRCh38
rs570755357 Chr.14: 55352275-55352275 on Build GRCh38
rs753388274 Chr.14: 55352118-55352118 on Build GRCh38
rs777976350 Chr.14: 55352202-55352202 on Build GRCh38
rs377547174 Chr.14: 55352288-55352288 on Build GRCh38
rs138193304 Chr.14: 55352249-55352249 on Build GRCh38
rs778899920 Chr.14: 55352274-55352274 on Build GRCh38
rs752582600 Chr.14: 55352198-55352198 on Build GRCh38
rs372485384 Chr.14: 55352267-55352267 on Build GRCh38
rs570992846 Chr.14: 55352292-55352292 on Build GRCh38
rs746919657 Chr.14: 55352258-55352258 on Build GRCh38
rs756077791 Chr.14: 55352200-55352200 on Build GRCh38
rs768631951 Chr.14: 55352266-55352266 on Build GRCh38
rs112254408 Chr.14: 55352148-55352148 on Build GRCh38
rs141781589 Chr.14: 55352110-55352110 on Build GRCh38
rs761506690 Chr.14: 55352136-55352136 on Build GRCh38
rs755306285 Chr.14: 55352154-55352154 on Build GRCh38
rs760130316 Chr.14: 55352117-55352117 on Build GRCh38
rs530670367 Chr.14: 55352116-55352116 on Build GRCh38
rs761300524 Chr.14: 55352282-55352282 on Build GRCh38
rs745618901 Chr.14: 55352241-55352241 on Build GRCh38
rs980976462 Chr.14: 55352225-55352225 on Build GRCh38
rs764859601 Chr.14: 55352138-55352138 on Build GRCh38
rs781743849 Chr.14: 55352169-55352169 on Build GRCh38
rs927019918 Chr.14: 55352228-55352228 on Build GRCh38
rs771178382 Chr.14: 55352214-55352214 on Build GRCh38
rs200207283 Chr.14: 55352211-55352211 on Build GRCh38
rs779006830 Chr.14: 55352234-55352234 on Build GRCh38
rs146325881 Chr.14: 55352142-55352142 on Build GRCh38
rs771979310 Chr.14: 55352095-55352095 on Build GRCh38
rs753284118 Chr.14: 55352199-55352199 on Build GRCh38
rs951638467 Chr.14: 55352171-55352171 on Build GRCh38

View Full Product Details