Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87663176-87663431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMP1
Entrez Gene ID
1758
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
ARHP, ARHR, DMP-1
UniGene
Hs.652366
Gene Chromosome Location
Chr.4: 87650302-87664361 on Build GRCh38
Transcripts
NM_001079911.2 NM_004407.3 XM_011531705.2 XM_011531706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3606862 c.1453G>A p.E485K Chr.4: 87663231-87663231 on Build GRCh38
4918380 c.1515T>C p.D505D Chr.4: 87663293-87663293 on Build GRCh38
3373596 c.1514A>T p.D505V Chr.4: 87663292-87663292 on Build GRCh38
2957211 c.1465C>T p.R489W Chr.4: 87663243-87663243 on Build GRCh38
4135429 c.1439A>G p.Q480R Chr.4: 87663217-87663217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201149134 Chr.4: 87663284-87663284 on Build GRCh38
rs748412771 Chr.4: 87663282-87663282 on Build GRCh38
rs757441962 Chr.4: 87663266-87663266 on Build GRCh38
rs574215585 Chr.4: 87663226-87663226 on Build GRCh38
rs373096835 Chr.4: 87663271-87663271 on Build GRCh38
rs534492304 Chr.4: 87663311-87663311 on Build GRCh38
rs751766468 Chr.4: 87663244-87663244 on Build GRCh38
rs765723704 Chr.4: 87663219-87663219 on Build GRCh38
rs771691318 Chr.4: 87663335-87663335 on Build GRCh38
rs765769920 Chr.4: 87663347-87663347 on Build GRCh38
rs542706129 Chr.4: 87663357-87663357 on Build GRCh38
rs776998280 Chr.4: 87663203-87663203 on Build GRCh38
rs905252788 Chr.4: 87663236-87663236 on Build GRCh38
rs148156611 Chr.4: 87663211-87663211 on Build GRCh38
rs755124488 Chr.4: 87663243-87663244 on Build GRCh38
rs911866027 Chr.4: 87663340-87663340 on Build GRCh38
rs587776698 Chr.4: 87663263-87663269 on Build GRCh38
rs576346276 Chr.4: 87663399-87663399 on Build GRCh38
rs780117938 Chr.4: 87663306-87663306 on Build GRCh38
rs781354531 Chr.4: 87663276-87663276 on Build GRCh38
rs963086141 Chr.4: 87663397-87663397 on Build GRCh38
rs759920278 Chr.4: 87663344-87663344 on Build GRCh38
rs949379753 Chr.4: 87663218-87663218 on Build GRCh38
rs1043882695 Chr.4: 87663392-87663392 on Build GRCh38
rs986468886 Chr.4: 87663334-87663334 on Build GRCh38
rs200014918 Chr.4: 87663201-87663201 on Build GRCh38
rs377414504 Chr.4: 87663312-87663312 on Build GRCh38
rs376245118 Chr.4: 87663268-87663268 on Build GRCh38
rs561440615 Chr.4: 87663393-87663393 on Build GRCh38
rs886059687 Chr.4: 87663234-87663234 on Build GRCh38
rs191574196 Chr.4: 87663267-87663267 on Build GRCh38
rs141968248 Chr.4: 87663243-87663243 on Build GRCh38
rs747771988 Chr.4: 87663332-87663332 on Build GRCh38
rs776880303 Chr.4: 87663343-87663343 on Build GRCh38
rs756030027 Chr.4: 87663299-87663299 on Build GRCh38
rs886059688 Chr.4: 87663381-87663381 on Build GRCh38
rs1007614713 Chr.4: 87663384-87663384 on Build GRCh38
rs961836346 Chr.4: 87663272-87663272 on Build GRCh38
rs765734588 Chr.4: 87663236-87663239 on Build GRCh38
rs953264697 Chr.4: 87663331-87663331 on Build GRCh38
rs2627724 Chr.4: 87663355-87663355 on Build GRCh38
rs1053943262 Chr.4: 87663290-87663290 on Build GRCh38
rs372806204 Chr.4: 87663227-87663227 on Build GRCh38
rs954999572 Chr.4: 87663381-87663381 on Build GRCh38
rs536209200 Chr.4: 87663351-87663351 on Build GRCh38
rs75244647 Chr.4: 87663350-87663350 on Build GRCh38
rs146762807 Chr.4: 87663280-87663280 on Build GRCh38

View Full Product Details