Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235177861-235178134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID4B
Entrez Gene ID
51742
Gene Name
AT-rich interaction domain 4B
Gene Aliases
BCAA, BRCAA1, RBBP1L1, RBP1L1, SAP180
UniGene
Hs.575782
Gene Chromosome Location
Chr.1: 235166895-235328240 on Build GRCh38
Transcripts
NM_001206794.1 NM_016374.5 NM_031371.3 NR_135074.1 XM_006711781.2 XM_011544212.2 XM_017001468.1 XM_017001469.1 XM_017001470.1 XM_017001471.1 XM_017001472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5724629 c.3386C>T p.S1129L Chr.1: 235177862-235177862 on Build GRCh38
6452789 c.3338G>A p.C1113Y Chr.1: 235177910-235177910 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs7534107 Chr.1: 235178031-235178031 on Build GRCh38
rs760615204 Chr.1: 235177936-235177937 on Build GRCh38
rs917516224 Chr.1: 235178058-235178058 on Build GRCh38
rs553632185 Chr.1: 235177963-235177963 on Build GRCh38
rs1040943827 Chr.1: 235177960-235177960 on Build GRCh38
rs996646944 Chr.1: 235178025-235178025 on Build GRCh38
rs919341977 Chr.1: 235178033-235178033 on Build GRCh38
rs747116272 Chr.1: 235177915-235177915 on Build GRCh38
rs948942390 Chr.1: 235178085-235178085 on Build GRCh38
rs1016753795 Chr.1: 235178012-235178012 on Build GRCh38
rs891492053 Chr.1: 235177897-235177897 on Build GRCh38
rs199556870 Chr.1: 235177907-235177907 on Build GRCh38
rs1030017527 Chr.1: 235177980-235177980 on Build GRCh38
rs770307814 Chr.1: 235177945-235177945 on Build GRCh38
rs781766030 Chr.1: 235177940-235177940 on Build GRCh38
rs992822241 Chr.1: 235178050-235178052 on Build GRCh38
rs943935092 Chr.1: 235177923-235177923 on Build GRCh38
rs770445143 Chr.1: 235178052-235178052 on Build GRCh38
rs769149558 Chr.1: 235177939-235177939 on Build GRCh38
rs748486085 Chr.1: 235177942-235177942 on Build GRCh38
rs780177824 Chr.1: 235177887-235177887 on Build GRCh38
rs963459069 Chr.1: 235178013-235178013 on Build GRCh38
rs1006741878 Chr.1: 235177999-235177999 on Build GRCh38
rs868498719 Chr.1: 235177904-235177904 on Build GRCh38
rs956756716 Chr.1: 235178106-235178106 on Build GRCh38
rs1031404139 Chr.1: 235178099-235178099 on Build GRCh38
rs111876191 Chr.1: 235178079-235178079 on Build GRCh38
rs997860833 Chr.1: 235177973-235177973 on Build GRCh38
rs763972634 Chr.1: 235177938-235177939 on Build GRCh38
rs547197547 Chr.1: 235177987-235177987 on Build GRCh38
rs903934002 Chr.1: 235178045-235178045 on Build GRCh38
rs886812250 Chr.1: 235177884-235177884 on Build GRCh38
rs191077289 Chr.1: 235177995-235177995 on Build GRCh38

View Full Product Details