Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107618253-107618506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAP29
Entrez Gene ID
55973
Gene Name
B-cell receptor-associated protein 29
Gene Aliases
BAP29
UniGene
Hs.303787
Gene Chromosome Location
Chr.7: 107579977-107623317 on Build GRCh38
Transcripts
NM_001008405.2 NM_018844.3 NR_027830.1 XM_006716051.2 XM_011516406.2 XM_011516407.1 XM_011516408.2 XM_017012436.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3254804 c.911delT p.L306fs*1 Chr.7: 107618469-107618469 on Build GRCh38
3949715 c.930A>G p.I310M Chr.7: 107618488-107618488 on Build GRCh38
1083867 c.915T>G p.F305L Chr.7: 107618473-107618473 on Build GRCh38
1673435 c.917T>A p.L306* Chr.7: 107618475-107618475 on Build GRCh38
1083866 c.773C>T p.S258L Chr.7: 107618331-107618331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115835071 Chr.7: 107618398-107618398 on Build GRCh38
rs777790092 Chr.7: 107618296-107618296 on Build GRCh38
rs534905033 Chr.7: 107618460-107618460 on Build GRCh38
rs752840337 Chr.7: 107618468-107618469 on Build GRCh38
rs910990658 Chr.7: 107618408-107618408 on Build GRCh38
rs759912941 Chr.7: 107618379-107618379 on Build GRCh38
rs764122441 Chr.7: 107618390-107618390 on Build GRCh38
rs774922774 Chr.7: 107618377-107618377 on Build GRCh38
rs771712834 Chr.7: 107618326-107618326 on Build GRCh38
rs10262001 Chr.7: 107618411-107618411 on Build GRCh38
rs202191845 Chr.7: 107618384-107618384 on Build GRCh38
rs772976784 Chr.7: 107618322-107618322 on Build GRCh38
rs760858542 Chr.7: 107618458-107618458 on Build GRCh38
rs753680659 Chr.7: 107618297-107618297 on Build GRCh38
rs778732433 Chr.7: 107618300-107618300 on Build GRCh38
rs537657093 Chr.7: 107618416-107618416 on Build GRCh38
rs201233658 Chr.7: 107618465-107618465 on Build GRCh38
rs568621453 Chr.7: 107618415-107618415 on Build GRCh38
rs370245331 Chr.7: 107618315-107618315 on Build GRCh38
rs750906931 Chr.7: 107618397-107618397 on Build GRCh38
rs753935872 Chr.7: 107618391-107618391 on Build GRCh38
rs771550713 Chr.7: 107618369-107618369 on Build GRCh38
rs6958094 Chr.7: 107618462-107618462 on Build GRCh38
rs756090536 Chr.7: 107618285-107618285 on Build GRCh38
rs550938616 Chr.7: 107618453-107618453 on Build GRCh38
rs755211991 Chr.7: 107618312-107618313 on Build GRCh38
rs750227463 Chr.7: 107618404-107618404 on Build GRCh38
rs757190632 Chr.7: 107618298-107618298 on Build GRCh38
rs774941391 Chr.7: 107618443-107618443 on Build GRCh38
rs754067990 Chr.7: 107618313-107618313 on Build GRCh38
rs780623261 Chr.7: 107618418-107618418 on Build GRCh38
rs747689058 Chr.7: 107618424-107618424 on Build GRCh38
rs551843466 Chr.7: 107618332-107618332 on Build GRCh38
rs141644610 Chr.7: 107618403-107618403 on Build GRCh38
rs775895217 Chr.7: 107618454-107618454 on Build GRCh38
rs866599232 Chr.7: 107618480-107618480 on Build GRCh38
rs750333436 Chr.7: 107618464-107618464 on Build GRCh38
rs768009337 Chr.7: 107618335-107618335 on Build GRCh38
rs766185837 Chr.7: 107618470-107618470 on Build GRCh38
rs374467600 Chr.7: 107618383-107618383 on Build GRCh38
rs776173375 Chr.7: 107618344-107618344 on Build GRCh38
rs138972250 Chr.7: 107618451-107618451 on Build GRCh38
rs146893215 Chr.7: 107618459-107618459 on Build GRCh38
rs751312535 Chr.7: 107618471-107618471 on Build GRCh38
rs779709844 Chr.7: 107618414-107618414 on Build GRCh38
rs771569195 Chr.7: 107618442-107618442 on Build GRCh38
rs267601226 Chr.7: 107618331-107618331 on Build GRCh38
rs763417273 Chr.7: 107618347-107618347 on Build GRCh38

View Full Product Details