Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67452230-67452495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP4
Entrez Gene ID
57010
Gene Name
calcium binding protein 4
Gene Aliases
CRSD, CSNB2B
UniGene
Hs.143036
Gene Chromosome Location
Chr.11: 67452406-67461774 on Build GRCh38
Transcripts
NM_001300896.1 XM_017018025.1 NM_206997.1

Gene Symbol
GPR152
Entrez Gene ID
390212
Gene Name
G protein-coupled receptor 152
Gene Aliases
PGR5
UniGene
Hs.567997
Gene Chromosome Location
Chr.11: 67451301-67452729 on Build GRCh38
Transcripts
NM_001300896.1 XM_017018025.1 NM_206997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5899024 c.333G>A p.W111* Chr.11: 67452392-67452392 on Build GRCh38
1356428 c.351C>A p.S117S Chr.11: 67452374-67452374 on Build GRCh38
1356427 c.476C>T p.A159V Chr.11: 67452249-67452249 on Build GRCh38
3452306 c.474G>A p.L158L Chr.11: 67452251-67452251 on Build GRCh38
931092 c.411C>T p.H137H Chr.11: 67452314-67452314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191782000 Chr.11: 67452340-67452340 on Build GRCh38
rs117903569 Chr.11: 67452326-67452326 on Build GRCh38
rs527749058 Chr.11: 67452266-67452266 on Build GRCh38
rs756679563 Chr.11: 67452277-67452277 on Build GRCh38
rs558662013 Chr.11: 67452450-67452450 on Build GRCh38
rs554577764 Chr.11: 67452344-67452344 on Build GRCh38
rs767785078 Chr.11: 67452397-67452397 on Build GRCh38
rs201384962 Chr.11: 67452411-67452411 on Build GRCh38
rs144630243 Chr.11: 67452252-67452252 on Build GRCh38
rs774285725 Chr.11: 67452389-67452389 on Build GRCh38
rs766828778 Chr.11: 67452412-67452412 on Build GRCh38
rs150873557 Chr.11: 67452475-67452475 on Build GRCh38
rs755426722 Chr.11: 67452432-67452432 on Build GRCh38
rs372634169 Chr.11: 67452355-67452355 on Build GRCh38
rs763037504 Chr.11: 67452323-67452323 on Build GRCh38
rs769833209 Chr.11: 67452383-67452383 on Build GRCh38
rs780722780 Chr.11: 67452279-67452279 on Build GRCh38
rs375896674 Chr.11: 67452441-67452441 on Build GRCh38
rs778580128 Chr.11: 67452293-67452293 on Build GRCh38
rs776535111 Chr.11: 67452374-67452374 on Build GRCh38
rs778093692 Chr.11: 67452388-67452388 on Build GRCh38
rs371602409 Chr.11: 67452274-67452274 on Build GRCh38
rs747752250 Chr.11: 67452356-67452356 on Build GRCh38
rs777354030 Chr.11: 67452451-67452451 on Build GRCh38
rs796337570 Chr.11: 67452276-67452277 on Build GRCh38
rs768083274 Chr.11: 67452291-67452291 on Build GRCh38
rs1013487922 Chr.11: 67452393-67452393 on Build GRCh38
rs757544766 Chr.11: 67452269-67452269 on Build GRCh38
rs780669506 Chr.11: 67452343-67452343 on Build GRCh38
rs748564423 Chr.11: 67452267-67452267 on Build GRCh38
rs367720617 Chr.11: 67452327-67452327 on Build GRCh38
rs746764243 Chr.11: 67452370-67452370 on Build GRCh38
rs867805011 Chr.11: 67452447-67452447 on Build GRCh38
rs776578974 Chr.11: 67452318-67452318 on Build GRCh38
rs560127653 Chr.11: 67452265-67452265 on Build GRCh38
rs866046742 Chr.11: 67452442-67452442 on Build GRCh38
rs145363353 Chr.11: 67452339-67452339 on Build GRCh38
rs745757748 Chr.11: 67452379-67452379 on Build GRCh38
rs533858336 Chr.11: 67452435-67452435 on Build GRCh38
rs758553167 Chr.11: 67452336-67452336 on Build GRCh38
rs554436929 Chr.11: 67452264-67452264 on Build GRCh38
rs750112029 Chr.11: 67452400-67452402 on Build GRCh38
rs760600715 Chr.11: 67452248-67452248 on Build GRCh38
rs752936476 Chr.11: 67452310-67452312 on Build GRCh38
rs138530126 Chr.11: 67452288-67452288 on Build GRCh38
rs780839937 Chr.11: 67452474-67452474 on Build GRCh38
rs755343622 Chr.11: 67452276-67452277 on Build GRCh38
rs376157455 Chr.11: 67452431-67452431 on Build GRCh38
rs771653529 Chr.11: 67452364-67452364 on Build GRCh38
rs200792057 Chr.11: 67452298-67452298 on Build GRCh38
rs777405785 Chr.11: 67452368-67452368 on Build GRCh38
rs376409557 Chr.11: 67452268-67452268 on Build GRCh38
rs761970403 Chr.11: 67452391-67452391 on Build GRCh38
rs760475740 Chr.11: 67452308-67452308 on Build GRCh38
rs1024941998 Chr.11: 67452395-67452395 on Build GRCh38
rs1002268917 Chr.11: 67452439-67452439 on Build GRCh38
rs773602358 Chr.11: 67452405-67452405 on Build GRCh38
rs746639392 Chr.11: 67452459-67452459 on Build GRCh38
rs957375863 Chr.11: 67452315-67452315 on Build GRCh38
rs1055556848 Chr.11: 67452385-67452385 on Build GRCh38
rs374518560 Chr.11: 67452295-67452295 on Build GRCh38
rs141536246 Chr.11: 67452473-67452473 on Build GRCh38
rs771778277 Chr.11: 67452297-67452297 on Build GRCh38
rs148000171 Chr.11: 67452373-67452373 on Build GRCh38
rs770786693 Chr.11: 67452314-67452314 on Build GRCh38

View Full Product Details