Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 39229174-39229429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL4
Entrez Gene ID
344387
Gene Name
cyclin dependent kinase like 4
Gene Aliases
-
UniGene
Hs.403201
Gene Chromosome Location
Chr.2: 39174421-39246815 on Build GRCh38
Transcripts
NM_001009565.1 XM_011532815.2 XM_011532816.2 XM_011532817.2 XM_011532819.2 XM_017003981.1 XM_017003982.1 XM_017003983.1 XM_017003984.1 XM_017003985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7360684 c.158G>A p.R53H Chr.2: 39229375-39229375 on Build GRCh38
5433898 c.141A>C p.I47I Chr.2: 39229392-39229392 on Build GRCh38
4869476 c.156A>G p.I52M Chr.2: 39229377-39229377 on Build GRCh38
3839526 c.115G>C p.E39Q Chr.2: 39229418-39229418 on Build GRCh38
6408167 c.130G>A p.V44I Chr.2: 39229403-39229403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934289603 Chr.2: 39229243-39229243 on Build GRCh38
rs938016110 Chr.2: 39229268-39229268 on Build GRCh38
rs753000622 Chr.2: 39229327-39229327 on Build GRCh38
rs754737936 Chr.2: 39229400-39229400 on Build GRCh38
rs772377501 Chr.2: 39229344-39229349 on Build GRCh38
rs983282232 Chr.2: 39229208-39229208 on Build GRCh38
rs116112545 Chr.2: 39229279-39229279 on Build GRCh38
rs924389610 Chr.2: 39229240-39229245 on Build GRCh38
rs551082412 Chr.2: 39229286-39229286 on Build GRCh38
rs568272885 Chr.2: 39229343-39229344 on Build GRCh38
rs962803513 Chr.2: 39229216-39229216 on Build GRCh38
rs757443116 Chr.2: 39229360-39229360 on Build GRCh38
rs764422054 Chr.2: 39229338-39229338 on Build GRCh38
rs143668771 Chr.2: 39229382-39229382 on Build GRCh38
rs35454041 Chr.2: 39229375-39229375 on Build GRCh38
rs765358203 Chr.2: 39229362-39229362 on Build GRCh38
rs1003948933 Chr.2: 39229344-39229345 on Build GRCh38
rs926630161 Chr.2: 39229253-39229253 on Build GRCh38
rs1051474325 Chr.2: 39229246-39229246 on Build GRCh38
rs766021428 Chr.2: 39229376-39229376 on Build GRCh38
rs554775197 Chr.2: 39229217-39229217 on Build GRCh38
rs761971067 Chr.2: 39229271-39229271 on Build GRCh38
rs776931063 Chr.2: 39229226-39229226 on Build GRCh38
rs754046232 Chr.2: 39229350-39229350 on Build GRCh38
rs35121800 Chr.2: 39229278-39229279 on Build GRCh38
rs746764745 Chr.2: 39229398-39229398 on Build GRCh38
rs974575506 Chr.2: 39229239-39229239 on Build GRCh38
rs573634459 Chr.2: 39229277-39229277 on Build GRCh38
rs569078757 Chr.2: 39229296-39229296 on Build GRCh38
rs183499483 Chr.2: 39229212-39229212 on Build GRCh38
rs761017908 Chr.2: 39229331-39229331 on Build GRCh38

View Full Product Details