Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 91976932-91977201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
UniGene
Hs.482520
Gene Chromosome Location
Chr.1: 91949371-92014428 on Build GRCh38
Transcripts
NM_001242805.2 NM_001242806.2 NM_001242807.2 NM_001242808.2 NM_001242810.2 NM_001726.4 NM_207189.3 XM_006710853.3 XM_006710854.3 XM_006710855.3 XM_006710856.3 XM_006710857.3 XM_011542032.2 XM_011542033.2 XM_011542034.2 XM_011542035.2 XM_011542036.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6196877 c.762G>T p.L254F Chr.1: 91977186-91977186 on Build GRCh38
4010587 c.731C>A p.P244H Chr.1: 91977155-91977155 on Build GRCh38
6612780 c.753G>T p.K251N Chr.1: 91977177-91977177 on Build GRCh38
2259835 c.753G>A p.K251K Chr.1: 91977177-91977177 on Build GRCh38
913229 c.736A>C p.K246Q Chr.1: 91977160-91977160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952910612 Chr.1: 91977103-91977103 on Build GRCh38
rs368897015 Chr.1: 91977087-91977087 on Build GRCh38
rs985111493 Chr.1: 91977016-91977016 on Build GRCh38
rs751684226 Chr.1: 91977137-91977137 on Build GRCh38
rs766798078 Chr.1: 91977159-91977159 on Build GRCh38
rs780558202 Chr.1: 91977039-91977039 on Build GRCh38
rs748093699 Chr.1: 91977018-91977020 on Build GRCh38
rs766037028 Chr.1: 91976995-91976995 on Build GRCh38
rs993105491 Chr.1: 91977145-91977145 on Build GRCh38
rs142020784 Chr.1: 91977125-91977125 on Build GRCh38
rs771728285 Chr.1: 91977074-91977074 on Build GRCh38
rs374654048 Chr.1: 91977023-91977023 on Build GRCh38
rs757539477 Chr.1: 91976998-91976998 on Build GRCh38
rs768083673 Chr.1: 91977055-91977055 on Build GRCh38
rs367765022 Chr.1: 91977026-91977026 on Build GRCh38
rs192529642 Chr.1: 91977143-91977143 on Build GRCh38
rs763138357 Chr.1: 91977122-91977122 on Build GRCh38
rs756459153 Chr.1: 91977158-91977158 on Build GRCh38
rs1156281 Chr.1: 91977138-91977138 on Build GRCh38
rs953705983 Chr.1: 91976969-91976970 on Build GRCh38
rs374042055 Chr.1: 91977171-91977171 on Build GRCh38
rs961945096 Chr.1: 91976980-91976980 on Build GRCh38
rs925717161 Chr.1: 91977060-91977060 on Build GRCh38
rs770805956 Chr.1: 91977105-91977105 on Build GRCh38
rs778546450 Chr.1: 91977065-91977065 on Build GRCh38
rs769956174 Chr.1: 91977073-91977075 on Build GRCh38
rs143654874 Chr.1: 91977083-91977083 on Build GRCh38
rs750774812 Chr.1: 91977150-91977150 on Build GRCh38
rs748824658 Chr.1: 91977050-91977050 on Build GRCh38
rs754759998 Chr.1: 91976993-91976993 on Build GRCh38
rs536015282 Chr.1: 91976972-91976976 on Build GRCh38
rs267598766 Chr.1: 91977057-91977057 on Build GRCh38
rs747865586 Chr.1: 91977070-91977070 on Build GRCh38
rs781335800 Chr.1: 91977010-91977011 on Build GRCh38
rs756627393 Chr.1: 91977036-91977036 on Build GRCh38
rs760472427 Chr.1: 91977094-91977094 on Build GRCh38
rs549068065 Chr.1: 91977119-91977119 on Build GRCh38
rs984184721 Chr.1: 91977130-91977130 on Build GRCh38

View Full Product Details