Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89894759-89895022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA10
Entrez Gene ID
84694
Gene Name
gap junction protein alpha 10
Gene Aliases
CX62
UniGene
Hs.334499
Gene Chromosome Location
Chr.6: 89894469-89896100 on Build GRCh38
Transcripts
NM_032602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4546951 c.410G>A p.R137K Chr.6: 89894878-89894878 on Build GRCh38
3630529 c.468T>C p.T156T Chr.6: 89894936-89894936 on Build GRCh38
5683831 c.486G>T p.L162F Chr.6: 89894954-89894954 on Build GRCh38
3630527 c.320G>A p.R107K Chr.6: 89894788-89894788 on Build GRCh38
5603647 c.412T>C p.L138L Chr.6: 89894880-89894880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368294375 Chr.6: 89894976-89894976 on Build GRCh38
rs575951396 Chr.6: 89894986-89894986 on Build GRCh38
rs898446932 Chr.6: 89894862-89894862 on Build GRCh38
rs750627146 Chr.6: 89894903-89894903 on Build GRCh38
rs939908509 Chr.6: 89894840-89894840 on Build GRCh38
rs371750239 Chr.6: 89894967-89894967 on Build GRCh38
rs762819519 Chr.6: 89894860-89894860 on Build GRCh38
rs775524517 Chr.6: 89894800-89894800 on Build GRCh38
rs748993836 Chr.6: 89894828-89894828 on Build GRCh38
rs540684772 Chr.6: 89894931-89894931 on Build GRCh38
rs758447136 Chr.6: 89894796-89894796 on Build GRCh38
rs1024872179 Chr.6: 89894923-89894923 on Build GRCh38
rs140054168 Chr.6: 89894885-89894885 on Build GRCh38
rs752696468 Chr.6: 89894924-89894924 on Build GRCh38
rs574056749 Chr.6: 89894939-89894939 on Build GRCh38
rs774812208 Chr.6: 89894957-89894957 on Build GRCh38
rs766383813 Chr.6: 89894912-89894912 on Build GRCh38
rs756085406 Chr.6: 89894877-89894877 on Build GRCh38
rs764211796 Chr.6: 89894994-89894994 on Build GRCh38
rs765329772 Chr.6: 89894786-89894786 on Build GRCh38
rs762219947 Chr.6: 89894991-89894991 on Build GRCh38
rs554819302 Chr.6: 89894807-89894807 on Build GRCh38
rs199908427 Chr.6: 89894973-89894973 on Build GRCh38
rs919322501 Chr.6: 89894864-89894864 on Build GRCh38
rs771720539 Chr.6: 89894954-89894954 on Build GRCh38
rs770669564 Chr.6: 89894843-89894843 on Build GRCh38
rs889298610 Chr.6: 89894907-89894907 on Build GRCh38
rs114953447 Chr.6: 89894880-89894880 on Build GRCh38
rs761028588 Chr.6: 89894877-89894877 on Build GRCh38
rs774116939 Chr.6: 89894846-89894846 on Build GRCh38
rs745951035 Chr.6: 89894875-89894875 on Build GRCh38
rs765487975 Chr.6: 89894902-89894902 on Build GRCh38
rs569183914 Chr.6: 89894785-89894785 on Build GRCh38
rs867647028 Chr.6: 89894823-89894823 on Build GRCh38
rs745446516 Chr.6: 89894946-89894946 on Build GRCh38
rs199710744 Chr.6: 89894814-89894814 on Build GRCh38
rs757096349 Chr.6: 89894936-89894936 on Build GRCh38
rs755945621 Chr.6: 89894808-89894808 on Build GRCh38
rs771704005 Chr.6: 89894851-89894851 on Build GRCh38
rs750539392 Chr.6: 89894789-89894789 on Build GRCh38
rs745419913 Chr.6: 89894847-89894847 on Build GRCh38
rs762203193 Chr.6: 89894896-89894896 on Build GRCh38
rs150298953 Chr.6: 89894932-89894932 on Build GRCh38
rs758545153 Chr.6: 89894909-89894909 on Build GRCh38

View Full Product Details