Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64221948-64222189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf66
Entrez Gene ID
144577
Gene Name
chromosome 12 open reading frame 66
Gene Aliases
-
UniGene
Hs.444671
Gene Chromosome Location
Chr.12: 64186312-64222296 on Build GRCh38
Transcripts
NM_001300940.1 NM_001300941.1 NM_152440.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4606792 c.62T>G p.F21C Chr.12: 64222176-64222176 on Build GRCh38
6614262 c.159G>A p.S53S Chr.12: 64222079-64222079 on Build GRCh38
1991039 c.99T>C p.A33A Chr.12: 64222139-64222139 on Build GRCh38
1991038 c.173T>A p.L58* Chr.12: 64222065-64222065 on Build GRCh38
3931718 c.179A>C p.H60P Chr.12: 64222059-64222059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974744122 Chr.12: 64222038-64222038 on Build GRCh38
rs200046866 Chr.12: 64222031-64222031 on Build GRCh38
rs749765095 Chr.12: 64221983-64221983 on Build GRCh38
rs764363193 Chr.12: 64222060-64222060 on Build GRCh38
rs770417449 Chr.12: 64221977-64221977 on Build GRCh38
rs377159432 Chr.12: 64222061-64222061 on Build GRCh38
rs755361054 Chr.12: 64222097-64222097 on Build GRCh38
rs759484652 Chr.12: 64222011-64222011 on Build GRCh38
rs759001709 Chr.12: 64222068-64222068 on Build GRCh38
rs371158542 Chr.12: 64222092-64222092 on Build GRCh38
rs778419516 Chr.12: 64222159-64222159 on Build GRCh38
rs771314426 Chr.12: 64222091-64222092 on Build GRCh38
rs760502678 Chr.12: 64222054-64222054 on Build GRCh38
rs745767177 Chr.12: 64222166-64222166 on Build GRCh38
rs912549823 Chr.12: 64222167-64222167 on Build GRCh38
rs982780020 Chr.12: 64222165-64222165 on Build GRCh38
rs374893440 Chr.12: 64222100-64222100 on Build GRCh38
rs773759109 Chr.12: 64221982-64221983 on Build GRCh38
rs777481409 Chr.12: 64222127-64222127 on Build GRCh38
rs774280599 Chr.12: 64221989-64221989 on Build GRCh38
rs775295898 Chr.12: 64222039-64222039 on Build GRCh38
rs188275145 Chr.12: 64221990-64221990 on Build GRCh38
rs1055569228 Chr.12: 64221984-64221984 on Build GRCh38
rs887937489 Chr.12: 64222104-64222104 on Build GRCh38
rs756736430 Chr.12: 64222148-64222148 on Build GRCh38
rs769852745 Chr.12: 64221976-64221978 on Build GRCh38
rs200715336 Chr.12: 64222150-64222150 on Build GRCh38
rs377326868 Chr.12: 64222146-64222146 on Build GRCh38
rs201997850 Chr.12: 64222067-64222067 on Build GRCh38
rs369698955 Chr.12: 64222063-64222063 on Build GRCh38
rs377745337 Chr.12: 64222084-64222084 on Build GRCh38
rs771968044 Chr.12: 64222168-64222168 on Build GRCh38
rs774647515 Chr.12: 64222104-64222106 on Build GRCh38

View Full Product Details