Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44242321-44242556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOPAZ1
Entrez Gene ID
375337
Gene Name
testis and ovary specific PAZ domain containing 1
Gene Aliases
C3orf77
UniGene
Hs.132125
Gene Chromosome Location
Chr.3: 44241886-44337071 on Build GRCh38
Transcripts
NM_001145030.1 XM_011533694.2 XM_017006361.1 XM_017006362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702435 c.346A>C p.T116P Chr.3: 44242399-44242399 on Build GRCh38
3066780 c.291G>A p.P97P Chr.3: 44242344-44242344 on Build GRCh38
1198828 c.269G>C p.S90T Chr.3: 44242322-44242322 on Build GRCh38
1044910 c.329T>C p.L110S Chr.3: 44242382-44242382 on Build GRCh38
4543442 c.337G>A p.E113K Chr.3: 44242390-44242390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902061540 Chr.3: 44242372-44242372 on Build GRCh38
rs546883406 Chr.3: 44242419-44242419 on Build GRCh38
rs533393196 Chr.3: 44242399-44242399 on Build GRCh38
rs1011729767 Chr.3: 44242415-44242415 on Build GRCh38
rs760958115 Chr.3: 44242410-44242410 on Build GRCh38
rs375336529 Chr.3: 44242381-44242381 on Build GRCh38
rs550279740 Chr.3: 44242443-44242443 on Build GRCh38
rs780989319 Chr.3: 44242452-44242452 on Build GRCh38
rs868617506 Chr.3: 44242368-44242368 on Build GRCh38
rs1049172963 Chr.3: 44242343-44242343 on Build GRCh38
rs184664960 Chr.3: 44242352-44242352 on Build GRCh38
rs13314833 Chr.3: 44242351-44242351 on Build GRCh38
rs762900653 Chr.3: 44242395-44242395 on Build GRCh38
rs370275967 Chr.3: 44242512-44242512 on Build GRCh38
rs776633930 Chr.3: 44242346-44242346 on Build GRCh38
rs928299589 Chr.3: 44242483-44242483 on Build GRCh38
rs1022748184 Chr.3: 44242432-44242432 on Build GRCh38
rs566284505 Chr.3: 44242422-44242422 on Build GRCh38
rs970126257 Chr.3: 44242454-44242454 on Build GRCh38
rs949944035 Chr.3: 44242373-44242373 on Build GRCh38
rs919746083 Chr.3: 44242521-44242521 on Build GRCh38
rs1031819738 Chr.3: 44242393-44242393 on Build GRCh38
rs548965778 Chr.3: 44242468-44242468 on Build GRCh38
rs767600568 Chr.3: 44242398-44242398 on Build GRCh38
rs1040779864 Chr.3: 44242359-44242359 on Build GRCh38
rs915803654 Chr.3: 44242369-44242369 on Build GRCh38
rs751189829 Chr.3: 44242388-44242388 on Build GRCh38
rs972644195 Chr.3: 44242514-44242514 on Build GRCh38
rs749872956 Chr.3: 44242341-44242341 on Build GRCh38
rs145719527 Chr.3: 44242456-44242456 on Build GRCh38
rs773054891 Chr.3: 44242374-44242374 on Build GRCh38

View Full Product Details