Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6271297-6271532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCKBR
Entrez Gene ID
887
Gene Name
cholecystokinin B receptor
Gene Aliases
CCK-B, CCK2R, GASR
UniGene
Hs.203
Gene Chromosome Location
Chr.11: 6259611-6272127 on Build GRCh38
Transcripts
NM_001318029.1 NM_176875.3 XM_005253210.1 XM_017018516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7202095 c.1399C>A p.R467S Chr.11: 6271391-6271391 on Build GRCh38
6551685 c.1229C>T p.P410L Chr.11: 6271428-6271428 on Build GRCh38
3356226 c.1423G>A p.A475T Chr.11: 6271415-6271415 on Build GRCh38
4463521 c.1295C>T p.S432L Chr.11: 6271494-6271494 on Build GRCh38
6622810 c.1239A>G p.P413P Chr.11: 6271438-6271438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757730115 Chr.11: 6271447-6271447 on Build GRCh38
rs767776959 Chr.11: 6271485-6271485 on Build GRCh38
rs772927840 Chr.11: 6271474-6271474 on Build GRCh38
rs201461045 Chr.11: 6271473-6271473 on Build GRCh38
rs200268499 Chr.11: 6271410-6271410 on Build GRCh38
rs555585232 Chr.11: 6271484-6271484 on Build GRCh38
rs775612982 Chr.11: 6271487-6271487 on Build GRCh38
rs202056290 Chr.11: 6271442-6271442 on Build GRCh38
rs749830885 Chr.11: 6271405-6271405 on Build GRCh38
rs753116644 Chr.11: 6271386-6271386 on Build GRCh38
rs201876764 Chr.11: 6271347-6271347 on Build GRCh38
rs753026530 Chr.11: 6271445-6271445 on Build GRCh38
rs773055083 Chr.11: 6271430-6271430 on Build GRCh38
rs760363464 Chr.11: 6271318-6271318 on Build GRCh38
rs533838696 Chr.11: 6271480-6271480 on Build GRCh38
rs145637819 Chr.11: 6271327-6271327 on Build GRCh38
rs766435444 Chr.11: 6271359-6271359 on Build GRCh38
rs773909097 Chr.11: 6271414-6271417 on Build GRCh38
rs200480495 Chr.11: 6271383-6271383 on Build GRCh38
rs199586552 Chr.11: 6271435-6271435 on Build GRCh38
rs76115904 Chr.11: 6271409-6271409 on Build GRCh38
rs201721347 Chr.11: 6271385-6271385 on Build GRCh38
rs200083878 Chr.11: 6271495-6271495 on Build GRCh38
rs138218747 Chr.11: 6271339-6271339 on Build GRCh38
rs755275816 Chr.11: 6271380-6271380 on Build GRCh38
rs764410444 Chr.11: 6271446-6271446 on Build GRCh38
rs201338739 Chr.11: 6271362-6271362 on Build GRCh38
rs149293522 Chr.11: 6271448-6271448 on Build GRCh38
rs993321475 Chr.11: 6271400-6271400 on Build GRCh38
rs937985143 Chr.11: 6271338-6271338 on Build GRCh38
rs756376172 Chr.11: 6271394-6271394 on Build GRCh38
rs897640161 Chr.11: 6271392-6271392 on Build GRCh38
rs749073181 Chr.11: 6271477-6271477 on Build GRCh38
rs573835273 Chr.11: 6271513-6271513 on Build GRCh38
rs748652241 Chr.11: 6271391-6271393 on Build GRCh38
rs200384464 Chr.11: 6271356-6271356 on Build GRCh38
rs765220669 Chr.11: 6271344-6271344 on Build GRCh38
rs1046862255 Chr.11: 6271348-6271348 on Build GRCh38
rs199726670 Chr.11: 6271415-6271415 on Build GRCh38
rs761678173 Chr.11: 6271427-6271427 on Build GRCh38
rs200837612 Chr.11: 6271426-6271426 on Build GRCh38
rs145064936 Chr.11: 6271397-6271397 on Build GRCh38
rs776196223 Chr.11: 6271421-6271421 on Build GRCh38
rs201366169 Chr.11: 6271414-6271414 on Build GRCh38
rs141279039 Chr.11: 6271357-6271357 on Build GRCh38
rs148059332 Chr.11: 6271494-6271494 on Build GRCh38
rs199896310 Chr.11: 6271331-6271331 on Build GRCh38
rs369943403 Chr.11: 6271436-6271436 on Build GRCh38
rs202203262 Chr.11: 6271376-6271376 on Build GRCh38
rs200777432 Chr.11: 6271470-6271470 on Build GRCh38
rs769534576 Chr.11: 6271428-6271428 on Build GRCh38
rs144569397 Chr.11: 6271464-6271464 on Build GRCh38
rs866773703 Chr.11: 6271439-6271439 on Build GRCh38
rs746523028 Chr.11: 6271463-6271463 on Build GRCh38
rs746470052 Chr.11: 6271413-6271413 on Build GRCh38
rs762933442 Chr.11: 6271358-6271358 on Build GRCh38
rs199498277 Chr.11: 6271408-6271408 on Build GRCh38
rs774373253 Chr.11: 6271479-6271479 on Build GRCh38
rs200728380 Chr.11: 6271440-6271440 on Build GRCh38
rs200919066 Chr.11: 6271340-6271340 on Build GRCh38
rs772679349 Chr.11: 6271396-6271404 on Build GRCh38
rs199904879 Chr.11: 6271468-6271468 on Build GRCh38
rs138829785 Chr.11: 6271431-6271431 on Build GRCh38

View Full Product Details