Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 22660826-22661089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QB
Entrez Gene ID
713
Gene Name
complement C1q B chain
Gene Aliases
-
UniGene
Hs.8986
Gene Chromosome Location
Chr.1: 22653189-22661536 on Build GRCh38
Transcripts
NM_000491.3 XM_011542059.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
242481 c.348C>A p.Y116* Chr.1: 22660972-22660972 on Build GRCh38
6124633 c.319C>A p.P107T Chr.1: 22660943-22660943 on Build GRCh38
905332 c.243G>A p.G81G Chr.1: 22660867-22660867 on Build GRCh38
6124631 c.406C>T p.R136W Chr.1: 22661030-22661030 on Build GRCh38
4880584 c.298G>T p.G100C Chr.1: 22660922-22660922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148746097 Chr.1: 22660957-22660957 on Build GRCh38
rs199696043 Chr.1: 22661031-22661031 on Build GRCh38
rs149072794 Chr.1: 22661030-22661030 on Build GRCh38
rs372067838 Chr.1: 22661046-22661046 on Build GRCh38
rs200457185 Chr.1: 22661018-22661018 on Build GRCh38
rs150683030 Chr.1: 22660998-22660998 on Build GRCh38
rs753074108 Chr.1: 22661028-22661028 on Build GRCh38
rs773232242 Chr.1: 22660880-22660880 on Build GRCh38
rs544508692 Chr.1: 22660892-22660892 on Build GRCh38
rs148813212 Chr.1: 22660941-22660941 on Build GRCh38
rs774367732 Chr.1: 22661020-22661020 on Build GRCh38
rs780155142 Chr.1: 22660922-22660929 on Build GRCh38
rs767893704 Chr.1: 22660934-22660934 on Build GRCh38
rs149771570 Chr.1: 22661004-22661004 on Build GRCh38
rs758439232 Chr.1: 22660909-22660909 on Build GRCh38
rs768600792 Chr.1: 22660866-22660866 on Build GRCh38
rs775542683 Chr.1: 22660854-22660854 on Build GRCh38
rs767948585 Chr.1: 22661023-22661023 on Build GRCh38
rs746836733 Chr.1: 22660858-22660858 on Build GRCh38
rs747942028 Chr.1: 22661051-22661051 on Build GRCh38
rs763074321 Chr.1: 22660882-22660882 on Build GRCh38
rs781175718 Chr.1: 22661050-22661050 on Build GRCh38
rs750117917 Chr.1: 22661039-22661039 on Build GRCh38
rs3211003 Chr.1: 22660884-22660884 on Build GRCh38
rs368280287 Chr.1: 22660891-22660891 on Build GRCh38
rs368860519 Chr.1: 22661064-22661064 on Build GRCh38
rs779720799 Chr.1: 22661044-22661044 on Build GRCh38
rs145412230 Chr.1: 22661017-22661017 on Build GRCh38
rs1056279304 Chr.1: 22661014-22661014 on Build GRCh38
rs745414763 Chr.1: 22660962-22660962 on Build GRCh38
rs753050059 Chr.1: 22660935-22660935 on Build GRCh38
rs778046227 Chr.1: 22660976-22660976 on Build GRCh38
rs776292843 Chr.1: 22660991-22660991 on Build GRCh38
rs749536477 Chr.1: 22661057-22661057 on Build GRCh38
rs867527641 Chr.1: 22660988-22660988 on Build GRCh38
rs776515186 Chr.1: 22660870-22660870 on Build GRCh38
rs200502843 Chr.1: 22660967-22660967 on Build GRCh38
rs376737563 Chr.1: 22661056-22661056 on Build GRCh38
rs755438683 Chr.1: 22660927-22660927 on Build GRCh38
rs908026459 Chr.1: 22660896-22660896 on Build GRCh38
rs757946445 Chr.1: 22660963-22660963 on Build GRCh38
rs751044051 Chr.1: 22661045-22661045 on Build GRCh38
rs1050307943 Chr.1: 22660939-22660939 on Build GRCh38
rs746963250 Chr.1: 22660990-22660990 on Build GRCh38
rs758073075 Chr.1: 22661040-22661040 on Build GRCh38
rs765432352 Chr.1: 22660879-22660879 on Build GRCh38
rs761952858 Chr.1: 22660872-22660872 on Build GRCh38
rs35477594 Chr.1: 22660847-22660847 on Build GRCh38
rs946701959 Chr.1: 22660948-22660948 on Build GRCh38
rs934072878 Chr.1: 22660923-22660923 on Build GRCh38

View Full Product Details