Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180529210-180529459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT6
Entrez Gene ID
57472
Gene Name
CCR4-NOT transcription complex subunit 6
Gene Aliases
CCR4, Ccr4a
UniGene
Hs.608380
Gene Chromosome Location
Chr.5: 180494373-180578405 on Build GRCh38
Transcripts
NM_001303241.1 XM_005265953.1 XM_011534605.2 XM_011534606.1 XM_011534607.1 XM_017009671.1 XM_017009672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3012324 c.55T>G p.S19A Chr.5: 180529331-180529331 on Build GRCh38
482645 c.80A>G p.K27R Chr.5: 180529356-180529356 on Build GRCh38
3919834 c.87C>T p.S29S Chr.5: 180529363-180529363 on Build GRCh38
4419509 c.26C>T p.P9L Chr.5: 180529302-180529302 on Build GRCh38
6222450 c.110G>A p.S37N Chr.5: 180529386-180529386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757566259 Chr.5: 180529241-180529241 on Build GRCh38
rs771961751 Chr.5: 180529250-180529250 on Build GRCh38
rs980270017 Chr.5: 180529282-180529282 on Build GRCh38
rs35618885 Chr.5: 180529266-180529267 on Build GRCh38
rs904885039 Chr.5: 180529423-180529423 on Build GRCh38
rs747693524 Chr.5: 180529370-180529370 on Build GRCh38
rs984749249 Chr.5: 180529259-180529259 on Build GRCh38
rs759938439 Chr.5: 180529293-180529293 on Build GRCh38
rs758027885 Chr.5: 180529414-180529414 on Build GRCh38
rs368230568 Chr.5: 180529375-180529375 on Build GRCh38
rs375788020 Chr.5: 180529248-180529248 on Build GRCh38
rs772817399 Chr.5: 180529281-180529281 on Build GRCh38
rs758646769 Chr.5: 180529325-180529325 on Build GRCh38
rs753922529 Chr.5: 180529304-180529304 on Build GRCh38
rs992933274 Chr.5: 180529328-180529328 on Build GRCh38
rs200597146 Chr.5: 180529311-180529311 on Build GRCh38
rs151053718 Chr.5: 180529342-180529342 on Build GRCh38
rs779318643 Chr.5: 180529320-180529320 on Build GRCh38
rs779746314 Chr.5: 180529333-180529333 on Build GRCh38
rs753062387 Chr.5: 180529297-180529297 on Build GRCh38
rs746944384 Chr.5: 180529256-180529256 on Build GRCh38
rs779263374 Chr.5: 180529246-180529246 on Build GRCh38
rs750652791 Chr.5: 180529324-180529324 on Build GRCh38
rs75964385 Chr.5: 180529302-180529302 on Build GRCh38
rs780090211 Chr.5: 180529251-180529251 on Build GRCh38
rs918625832 Chr.5: 180529336-180529336 on Build GRCh38
rs917635578 Chr.5: 180529316-180529316 on Build GRCh38
rs762623426 Chr.5: 180529404-180529404 on Build GRCh38
rs761090041 Chr.5: 180529300-180529300 on Build GRCh38
rs761258912 Chr.5: 180529266-180529266 on Build GRCh38
rs781179908 Chr.5: 180529359-180529359 on Build GRCh38
rs909147434 Chr.5: 180529276-180529276 on Build GRCh38
rs768601648 Chr.5: 180529258-180529258 on Build GRCh38
rs772657937 Chr.5: 180529387-180529387 on Build GRCh38
rs768494014 Chr.5: 180529343-180529343 on Build GRCh38

View Full Product Details