Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19788518-19788755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB1L
Entrez Gene ID
54584
Gene Name
G protein subunit beta 1 like
Gene Aliases
DGCRK3, FKSG1, GY2, WDR14, WDVCF
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19788411-19854939 on Build GRCh38
Transcripts
NM_053004.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1032138 c.972C>T p.Y324Y Chr.22: 19788721-19788721 on Build GRCh38
478730 c.980C>A p.A327E Chr.22: 19788713-19788713 on Build GRCh38
4562788 c.943G>A p.D315N Chr.22: 19788750-19788750 on Build GRCh38
7100921 c.979G>T p.A327S Chr.22: 19788714-19788714 on Build GRCh38
1682103 c.980C>T p.A327V Chr.22: 19788713-19788713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199731300 Chr.22: 19788633-19788634 on Build GRCh38
rs370115056 Chr.22: 19788621-19788621 on Build GRCh38
rs756031038 Chr.22: 19788674-19788674 on Build GRCh38
rs371580507 Chr.22: 19788605-19788605 on Build GRCh38
rs755330121 Chr.22: 19788664-19788664 on Build GRCh38
rs772618036 Chr.22: 19788687-19788687 on Build GRCh38
rs368471382 Chr.22: 19788580-19788580 on Build GRCh38
rs150948457 Chr.22: 19788717-19788717 on Build GRCh38
rs753964349 Chr.22: 19788623-19788623 on Build GRCh38
rs1012718511 Chr.22: 19788735-19788735 on Build GRCh38
rs925220096 Chr.22: 19788573-19788573 on Build GRCh38
rs376626625 Chr.22: 19788684-19788684 on Build GRCh38
rs112713709 Chr.22: 19788627-19788627 on Build GRCh38
rs374197241 Chr.22: 19788579-19788579 on Build GRCh38
rs759557690 Chr.22: 19788698-19788698 on Build GRCh38
rs768150223 Chr.22: 19788681-19788681 on Build GRCh38
rs767357362 Chr.22: 19788714-19788714 on Build GRCh38
rs374630161 Chr.22: 19788715-19788715 on Build GRCh38
rs3214094 Chr.22: 19788636-19788637 on Build GRCh38
rs773702352 Chr.22: 19788690-19788690 on Build GRCh38
rs919264953 Chr.22: 19788554-19788554 on Build GRCh38
rs943671687 Chr.22: 19788604-19788604 on Build GRCh38
rs376698385 Chr.22: 19788590-19788590 on Build GRCh38
rs766627090 Chr.22: 19788611-19788611 on Build GRCh38
rs777222184 Chr.22: 19788693-19788693 on Build GRCh38
rs779017197 Chr.22: 19788651-19788651 on Build GRCh38
rs77014742 Chr.22: 19788619-19788619 on Build GRCh38
rs574384524 Chr.22: 19788673-19788673 on Build GRCh38
rs899587591 Chr.22: 19788622-19788622 on Build GRCh38
rs752902364 Chr.22: 19788705-19788705 on Build GRCh38
rs545094749 Chr.22: 19788729-19788729 on Build GRCh38
rs142881986 Chr.22: 19788723-19788724 on Build GRCh38
rs761333857 Chr.22: 19788691-19788691 on Build GRCh38
rs760148141 Chr.22: 19788683-19788683 on Build GRCh38
rs972058095 Chr.22: 19788545-19788545 on Build GRCh38
rs894313261 Chr.22: 19788721-19788721 on Build GRCh38
rs7287799 Chr.22: 19788583-19788583 on Build GRCh38
rs758623156 Chr.22: 19788712-19788712 on Build GRCh38
rs779603312 Chr.22: 19788675-19788675 on Build GRCh38
rs751011522 Chr.22: 19788603-19788603 on Build GRCh38
rs528459018 Chr.22: 19788716-19788716 on Build GRCh38
rs3088076 Chr.22: 19788648-19788648 on Build GRCh38
rs72564428 Chr.22: 19788635-19788636 on Build GRCh38
rs757033818 Chr.22: 19788652-19788652 on Build GRCh38
rs765320708 Chr.22: 19788704-19788704 on Build GRCh38
rs979257623 Chr.22: 19788565-19788565 on Build GRCh38
rs767000148 Chr.22: 19788692-19788692 on Build GRCh38
rs1027144084 Chr.22: 19788655-19788655 on Build GRCh38
rs543298236 Chr.22: 19788680-19788680 on Build GRCh38
rs780932967 Chr.22: 19788661-19788661 on Build GRCh38
rs755020816 Chr.22: 19788636-19788636 on Build GRCh38

View Full Product Details