Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138467137-138467376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR1
Entrez Gene ID
1958
Gene Name
early growth response 1
Gene Aliases
AT225, G0S30, KROX-24, NGFI-A, TIS8, ZIF-268, ZNF225
UniGene
Hs.326035
Gene Chromosome Location
Chr.5: 138465492-138469315 on Build GRCh38
Transcripts
NM_001964.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5958356 c.877_878insAGG p.K293_A294insE Chr.5: 138467326-138467327 on Build GRCh38
5930246 c.762C>A p.D254E Chr.5: 138467211-138467211 on Build GRCh38
6652325 c.919C>A p.L307I Chr.5: 138467368-138467368 on Build GRCh38
1061390 c.839G>A p.R280H Chr.5: 138467288-138467288 on Build GRCh38
6652323 c.897G>A p.S299S Chr.5: 138467346-138467346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750817741 Chr.5: 138467268-138467268 on Build GRCh38
rs768039672 Chr.5: 138467262-138467262 on Build GRCh38
rs780315115 Chr.5: 138467176-138467176 on Build GRCh38
rs376540800 Chr.5: 138467215-138467215 on Build GRCh38
rs778893817 Chr.5: 138467195-138467195 on Build GRCh38
rs771996047 Chr.5: 138467211-138467211 on Build GRCh38
rs777476660 Chr.5: 138467212-138467212 on Build GRCh38
rs200832114 Chr.5: 138467222-138467222 on Build GRCh38
rs200009922 Chr.5: 138467229-138467229 on Build GRCh38
rs752798429 Chr.5: 138467304-138467304 on Build GRCh38
rs1030067845 Chr.5: 138467169-138467169 on Build GRCh38
rs969339195 Chr.5: 138467321-138467321 on Build GRCh38
rs975622774 Chr.5: 138467280-138467280 on Build GRCh38
rs201835292 Chr.5: 138467250-138467250 on Build GRCh38
rs748115078 Chr.5: 138467208-138467208 on Build GRCh38
rs982105424 Chr.5: 138467331-138467331 on Build GRCh38
rs777328073 Chr.5: 138467256-138467257 on Build GRCh38
rs199646874 Chr.5: 138467255-138467255 on Build GRCh38
rs199760996 Chr.5: 138467157-138467157 on Build GRCh38
rs753974749 Chr.5: 138467181-138467181 on Build GRCh38
rs568294551 Chr.5: 138467285-138467285 on Build GRCh38
rs201493880 Chr.5: 138467338-138467338 on Build GRCh38
rs755152311 Chr.5: 138467291-138467291 on Build GRCh38
rs770548315 Chr.5: 138467221-138467221 on Build GRCh38
rs200056344 Chr.5: 138467299-138467299 on Build GRCh38
rs775094635 Chr.5: 138467252-138467252 on Build GRCh38
rs762279773 Chr.5: 138467257-138467257 on Build GRCh38
rs375123090 Chr.5: 138467232-138467232 on Build GRCh38
rs755060107 Chr.5: 138467185-138467185 on Build GRCh38
rs201877780 Chr.5: 138467292-138467292 on Build GRCh38
rs756333418 Chr.5: 138467175-138467175 on Build GRCh38
rs141735308 Chr.5: 138467346-138467346 on Build GRCh38
rs767798551 Chr.5: 138467156-138467156 on Build GRCh38
rs759198636 Chr.5: 138467236-138467236 on Build GRCh38
rs865781599 Chr.5: 138467258-138467258 on Build GRCh38
rs200710842 Chr.5: 138467286-138467286 on Build GRCh38
rs866895287 Chr.5: 138467275-138467275 on Build GRCh38
rs868815253 Chr.5: 138467297-138467297 on Build GRCh38
rs201032646 Chr.5: 138467336-138467336 on Build GRCh38
rs974073836 Chr.5: 138467272-138467272 on Build GRCh38

View Full Product Details