Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45700169-45700426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN10
Entrez Gene ID
25814
Gene Name
ataxin 10
Gene Aliases
E46L, HUMEEP, SCA10
UniGene
Hs.475125
Gene Chromosome Location
Chr.22: 45671798-45845307 on Build GRCh38
Transcripts
NM_001167621.1 NM_013236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3912989 c.397C>T p.R133C Chr.22: 45700287-45700287 on Build GRCh38
1416820 c.433C>T p.R145W Chr.22: 45700323-45700323 on Build GRCh38
6608680 c.447C>T p.S149S Chr.22: 45700337-45700337 on Build GRCh38
4664650 c.460T>C p.W154R Chr.22: 45700350-45700350 on Build GRCh38
4104802 c.393T>C p.A131A Chr.22: 45700283-45700283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762802128 Chr.22: 45700369-45700369 on Build GRCh38
rs967264716 Chr.22: 45700245-45700245 on Build GRCh38
rs561324432 Chr.22: 45700344-45700344 on Build GRCh38
rs766403662 Chr.22: 45700287-45700287 on Build GRCh38
rs1033982518 Chr.22: 45700222-45700222 on Build GRCh38
rs766757216 Chr.22: 45700265-45700266 on Build GRCh38
rs751612343 Chr.22: 45700288-45700288 on Build GRCh38
rs777307727 Chr.22: 45700303-45700303 on Build GRCh38
rs767126969 Chr.22: 45700238-45700238 on Build GRCh38
rs760904363 Chr.22: 45700235-45700235 on Build GRCh38
rs777821087 Chr.22: 45700244-45700245 on Build GRCh38
rs369244529 Chr.22: 45700301-45700301 on Build GRCh38
rs142611714 Chr.22: 45700296-45700296 on Build GRCh38
rs752166502 Chr.22: 45700299-45700299 on Build GRCh38
rs199766375 Chr.22: 45700294-45700294 on Build GRCh38
rs747063252 Chr.22: 45700357-45700357 on Build GRCh38
rs768863649 Chr.22: 45700366-45700366 on Build GRCh38
rs754253984 Chr.22: 45700292-45700293 on Build GRCh38
rs757688241 Chr.22: 45700324-45700324 on Build GRCh38
rs371150803 Chr.22: 45700397-45700397 on Build GRCh38
rs759834291 Chr.22: 45700387-45700388 on Build GRCh38
rs957277745 Chr.22: 45700223-45700223 on Build GRCh38
rs34652509 Chr.22: 45700296-45700297 on Build GRCh38
rs545336515 Chr.22: 45700269-45700269 on Build GRCh38
rs776773929 Chr.22: 45700367-45700367 on Build GRCh38
rs140406039 Chr.22: 45700336-45700336 on Build GRCh38
rs773641734 Chr.22: 45700245-45700248 on Build GRCh38
rs774405755 Chr.22: 45700376-45700376 on Build GRCh38
rs780067797 Chr.22: 45700348-45700348 on Build GRCh38
rs770888323 Chr.22: 45700371-45700371 on Build GRCh38
rs541529533 Chr.22: 45700277-45700277 on Build GRCh38
rs746888231 Chr.22: 45700353-45700353 on Build GRCh38
rs764255013 Chr.22: 45700242-45700242 on Build GRCh38
rs746424807 Chr.22: 45700338-45700338 on Build GRCh38
rs150548393 Chr.22: 45700323-45700323 on Build GRCh38
rs1001236344 Chr.22: 45700203-45700203 on Build GRCh38

View Full Product Details