Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53270542-53270803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL5
Entrez Gene ID
60481
Gene Name
ELOVL fatty acid elongase 5
Gene Aliases
HELO1, SCA38, dJ483K16.1
UniGene
Hs.520189
Gene Chromosome Location
Chr.6: 53267398-53349179 on Build GRCh38
Transcripts
NM_001242828.1 NM_001242830.1 NM_001301856.1 NM_021814.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
484208 c.640A>G p.I214V Chr.6: 53270709-53270709 on Build GRCh38
5915529 c.756+3A>G p.? Chr.6: 53270590-53270590 on Build GRCh38
6283228 c.743A>C p.N248T Chr.6: 53270606-53270606 on Build GRCh38
5915531 c.622-7C>T p.? Chr.6: 53270734-53270734 on Build GRCh38
5915528 c.837+3A>G p.? Chr.6: 53270590-53270590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757916311 Chr.6: 53270772-53270772 on Build GRCh38
rs771472574 Chr.6: 53270743-53270743 on Build GRCh38
rs370251560 Chr.6: 53270731-53270731 on Build GRCh38
rs763841620 Chr.6: 53270678-53270678 on Build GRCh38
rs373426093 Chr.6: 53270659-53270659 on Build GRCh38
rs767915953 Chr.6: 53270751-53270751 on Build GRCh38
rs911034986 Chr.6: 53270741-53270741 on Build GRCh38
rs773445278 Chr.6: 53270734-53270734 on Build GRCh38
rs768175361 Chr.6: 53270753-53270753 on Build GRCh38
rs200269657 Chr.6: 53270677-53270677 on Build GRCh38
rs759926731 Chr.6: 53270750-53270750 on Build GRCh38
rs748684094 Chr.6: 53270653-53270656 on Build GRCh38
rs115953222 Chr.6: 53270739-53270739 on Build GRCh38
rs1010981524 Chr.6: 53270602-53270602 on Build GRCh38
rs760178808 Chr.6: 53270611-53270611 on Build GRCh38
rs779651994 Chr.6: 53270673-53270673 on Build GRCh38
rs893055134 Chr.6: 53270780-53270780 on Build GRCh38
rs760044461 Chr.6: 53270707-53270707 on Build GRCh38
rs587777670 Chr.6: 53270660-53270660 on Build GRCh38
rs193241282 Chr.6: 53270753-53270753 on Build GRCh38
rs373489693 Chr.6: 53270691-53270691 on Build GRCh38
rs528789078 Chr.6: 53270758-53270758 on Build GRCh38
rs980858218 Chr.6: 53270760-53270760 on Build GRCh38
rs774783374 Chr.6: 53270746-53270746 on Build GRCh38
rs41273880 Chr.6: 53270651-53270651 on Build GRCh38
rs201557998 Chr.6: 53270692-53270692 on Build GRCh38
rs751501083 Chr.6: 53270774-53270774 on Build GRCh38
rs756735815 Chr.6: 53270653-53270653 on Build GRCh38
rs959463552 Chr.6: 53270566-53270566 on Build GRCh38
rs140509241 Chr.6: 53270773-53270773 on Build GRCh38
rs373986125 Chr.6: 53270767-53270767 on Build GRCh38
rs941144260 Chr.6: 53270765-53270765 on Build GRCh38
rs200213620 Chr.6: 53270709-53270709 on Build GRCh38
rs749958366 Chr.6: 53270771-53270771 on Build GRCh38
rs763728017 Chr.6: 53270631-53270631 on Build GRCh38
rs111658712 Chr.6: 53270770-53270770 on Build GRCh38
rs754772768 Chr.6: 53270778-53270778 on Build GRCh38
rs753775748 Chr.6: 53270701-53270701 on Build GRCh38

View Full Product Details