Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28716778-28717045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL3
Entrez Gene ID
2137
Gene Name
exostosin like glycosyltransferase 3
Gene Aliases
BOTV, EXTL1L, EXTR1, REGR, RPR
UniGene
Hs.491354
Gene Chromosome Location
Chr.8: 28701473-28753690 on Build GRCh38
Transcripts
NM_001440.3 NR_073468.1 NR_073469.1 XM_011544440.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2788701 c.743C>T p.P248L Chr.8: 28716802-28716802 on Build GRCh38
1205871 c.780G>C p.L260F Chr.8: 28716839-28716839 on Build GRCh38
3899435 c.904A>G p.T302A Chr.8: 28716963-28716963 on Build GRCh38
5814649 c.720A>G p.I240M Chr.8: 28716779-28716779 on Build GRCh38
4644860 c.754C>T p.R252W Chr.8: 28716813-28716813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746487539 Chr.8: 28716992-28716992 on Build GRCh38
rs751017977 Chr.8: 28716853-28716853 on Build GRCh38
rs758298762 Chr.8: 28716988-28716988 on Build GRCh38
rs566700102 Chr.8: 28716816-28716816 on Build GRCh38
rs770324229 Chr.8: 28716932-28716932 on Build GRCh38
rs762271982 Chr.8: 28716851-28716851 on Build GRCh38
rs934606663 Chr.8: 28716947-28716947 on Build GRCh38
rs1012547826 Chr.8: 28716958-28716958 on Build GRCh38
rs199831414 Chr.8: 28716972-28716972 on Build GRCh38
rs762247666 Chr.8: 28716930-28716930 on Build GRCh38
rs149014454 Chr.8: 28717013-28717013 on Build GRCh38
rs369682689 Chr.8: 28716942-28716942 on Build GRCh38
rs774971340 Chr.8: 28716833-28716833 on Build GRCh38
rs768072663 Chr.8: 28716993-28716993 on Build GRCh38
rs779672202 Chr.8: 28716907-28716907 on Build GRCh38
rs757011891 Chr.8: 28716980-28716980 on Build GRCh38
rs768888582 Chr.8: 28716840-28716840 on Build GRCh38
rs36092162 Chr.8: 28716929-28716929 on Build GRCh38
rs1037401037 Chr.8: 28716893-28716893 on Build GRCh38
rs145734714 Chr.8: 28716897-28716897 on Build GRCh38
rs373044711 Chr.8: 28716859-28716859 on Build GRCh38
rs546457214 Chr.8: 28717005-28717005 on Build GRCh38
rs948845467 Chr.8: 28716860-28716860 on Build GRCh38
rs751595051 Chr.8: 28716956-28716956 on Build GRCh38
rs200720285 Chr.8: 28716986-28716986 on Build GRCh38
rs199663088 Chr.8: 28716889-28716889 on Build GRCh38
rs892795315 Chr.8: 28716951-28716951 on Build GRCh38
rs17059326 Chr.8: 28716863-28716863 on Build GRCh38
rs779860461 Chr.8: 28716990-28716990 on Build GRCh38
rs554162593 Chr.8: 28716813-28716813 on Build GRCh38
rs200615501 Chr.8: 28716862-28716862 on Build GRCh38
rs768286829 Chr.8: 28716925-28716925 on Build GRCh38
rs767597774 Chr.8: 28716999-28717029 on Build GRCh38
rs774306440 Chr.8: 28716811-28716811 on Build GRCh38
rs200303029 Chr.8: 28717004-28717004 on Build GRCh38
rs746513414 Chr.8: 28716913-28716913 on Build GRCh38
rs997196566 Chr.8: 28717011-28717011 on Build GRCh38
rs560087621 Chr.8: 28716974-28716974 on Build GRCh38
rs749621890 Chr.8: 28717012-28717012 on Build GRCh38
rs771658558 Chr.8: 28716814-28716814 on Build GRCh38
rs773851874 Chr.8: 28716941-28716941 on Build GRCh38
rs750343583 Chr.8: 28716987-28716987 on Build GRCh38
rs112390366 Chr.8: 28717016-28717016 on Build GRCh38
rs574279334 Chr.8: 28717022-28717022 on Build GRCh38
rs139633425 Chr.8: 28716898-28716898 on Build GRCh38
rs185956127 Chr.8: 28716810-28716810 on Build GRCh38
rs558506163 Chr.8: 28716858-28716858 on Build GRCh38
rs201627248 Chr.8: 28717001-28717001 on Build GRCh38
rs990390667 Chr.8: 28716835-28716835 on Build GRCh38
rs866103778 Chr.8: 28716950-28716950 on Build GRCh38
rs771474208 Chr.8: 28716899-28716899 on Build GRCh38
rs148151365 Chr.8: 28717017-28717017 on Build GRCh38
rs752962811 Chr.8: 28716979-28716979 on Build GRCh38
rs147630101 Chr.8: 28716943-28716943 on Build GRCh38
rs760437749 Chr.8: 28716839-28716839 on Build GRCh38
rs938712890 Chr.8: 28716885-28716885 on Build GRCh38
rs375285295 Chr.8: 28716802-28716802 on Build GRCh38
rs778742744 Chr.8: 28716983-28716983 on Build GRCh38
rs369906881 Chr.8: 28716803-28716803 on Build GRCh38
rs34985706 Chr.8: 28716846-28716846 on Build GRCh38

View Full Product Details