Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102918337-102918562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM2
Entrez Gene ID
54805
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 2
Gene Aliases
ACDP2, HOMG6, HOMGSMR
UniGene
Hs.643509
Gene Chromosome Location
Chr.10: 102918293-103087169 on Build GRCh38
Transcripts
NM_017649.4 NM_199076.2 NM_199077.2 XM_005269933.4 XM_006717908.2 XM_011539911.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6631308 c.50A>G p.Q17R Chr.10: 102918530-102918530 on Build GRCh38
5969629 c.16G>T p.A6S Chr.10: 102918496-102918496 on Build GRCh38
1321565 c.36G>C p.K12N Chr.10: 102918516-102918516 on Build GRCh38
401103 c.63A>T p.A21A Chr.10: 102918543-102918543 on Build GRCh38
1321564 c.36G>C p.K12N Chr.10: 102918516-102918516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576829658 Chr.10: 102918358-102918362 on Build GRCh38
rs931396715 Chr.10: 102918395-102918395 on Build GRCh38
rs868418989 Chr.10: 102918417-102918417 on Build GRCh38
rs769683854 Chr.10: 102918442-102918442 on Build GRCh38
rs745801073 Chr.10: 102918439-102918439 on Build GRCh38
rs200112200 Chr.10: 102918474-102918474 on Build GRCh38
rs147656969 Chr.10: 102918366-102918366 on Build GRCh38
rs1002524596 Chr.10: 102918383-102918383 on Build GRCh38
rs982593071 Chr.10: 102918536-102918536 on Build GRCh38
rs952539814 Chr.10: 102918494-102918494 on Build GRCh38
rs751242496 Chr.10: 102918431-102918431 on Build GRCh38
rs766846061 Chr.10: 102918542-102918542 on Build GRCh38
rs555951352 Chr.10: 102918371-102918371 on Build GRCh38
rs527368974 Chr.10: 102918398-102918398 on Build GRCh38
rs756689358 Chr.10: 102918433-102918433 on Build GRCh38
rs749083503 Chr.10: 102918471-102918471 on Build GRCh38
rs768606756 Chr.10: 102918480-102918480 on Build GRCh38
rs1048569859 Chr.10: 102918422-102918422 on Build GRCh38
rs772268353 Chr.10: 102918504-102918504 on Build GRCh38
rs574483645 Chr.10: 102918388-102918388 on Build GRCh38
rs1004886122 Chr.10: 102918463-102918463 on Build GRCh38
rs1046618439 Chr.10: 102918497-102918497 on Build GRCh38
rs562796364 Chr.10: 102918357-102918357 on Build GRCh38
rs774651042 Chr.10: 102918481-102918481 on Build GRCh38
rs1041971718 Chr.10: 102918364-102918364 on Build GRCh38
rs759044881 Chr.10: 102918409-102918409 on Build GRCh38
rs762143326 Chr.10: 102918492-102918492 on Build GRCh38
rs903428380 Chr.10: 102918368-102918368 on Build GRCh38
rs886046669 Chr.10: 102918529-102918529 on Build GRCh38
rs777218731 Chr.10: 102918543-102918543 on Build GRCh38
rs1027119969 Chr.10: 102918420-102918420 on Build GRCh38
rs370580602 Chr.10: 102918527-102918527 on Build GRCh38
rs886046668 Chr.10: 102918416-102918416 on Build GRCh38
rs761214082 Chr.10: 102918537-102918537 on Build GRCh38
rs559760130 Chr.10: 102918435-102918435 on Build GRCh38
rs779866075 Chr.10: 102918467-102918467 on Build GRCh38
rs921383268 Chr.10: 102918382-102918382 on Build GRCh38

View Full Product Details