Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55743704-55743929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF11
Entrez Gene ID
10220
Gene Name
growth differentiation factor 11
Gene Aliases
BMP-11, BMP11
UniGene
Hs.600883
Gene Chromosome Location
Chr.12: 55743278-55757194 on Build GRCh38
Transcripts
NM_005811.3 XM_006719194.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3463262 c.391G>A p.E131K Chr.12: 55743707-55743707 on Build GRCh38
3384506 c.426C>A p.V142V Chr.12: 55743742-55743742 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368915858 Chr.12: 55743734-55743734 on Build GRCh38
rs755767888 Chr.12: 55743748-55743748 on Build GRCh38
rs775708403 Chr.12: 55743793-55743793 on Build GRCh38
rs565621078 Chr.12: 55743859-55743859 on Build GRCh38
rs931469485 Chr.12: 55743889-55743889 on Build GRCh38
rs535365448 Chr.12: 55743805-55743805 on Build GRCh38
rs1015691591 Chr.12: 55743904-55743904 on Build GRCh38
rs763131962 Chr.12: 55743809-55743809 on Build GRCh38
rs769969074 Chr.12: 55743791-55743791 on Build GRCh38
rs374537273 Chr.12: 55743725-55743725 on Build GRCh38
rs529877011 Chr.12: 55743884-55743884 on Build GRCh38
rs778268404 Chr.12: 55743780-55743780 on Build GRCh38
rs748861162 Chr.12: 55743757-55743757 on Build GRCh38
rs765742555 Chr.12: 55743882-55743882 on Build GRCh38
rs970951334 Chr.12: 55743868-55743868 on Build GRCh38
rs757096616 Chr.12: 55743731-55743731 on Build GRCh38
rs758834283 Chr.12: 55743768-55743768 on Build GRCh38
rs78678275 Chr.12: 55743897-55743897 on Build GRCh38
rs746080564 Chr.12: 55743790-55743790 on Build GRCh38
rs750059741 Chr.12: 55743735-55743735 on Build GRCh38
rs747290113 Chr.12: 55743782-55743782 on Build GRCh38
rs776904830 Chr.12: 55743789-55743789 on Build GRCh38
rs372296030 Chr.12: 55743753-55743753 on Build GRCh38
rs879151292 Chr.12: 55743728-55743728 on Build GRCh38
rs771362049 Chr.12: 55743788-55743788 on Build GRCh38

View Full Product Details