Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153948984-153949249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC2
Entrez Gene ID
200186
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
TORC-2, TORC2
UniGene
Hs.406392
Gene Chromosome Location
Chr.1: 153947672-153958656 on Build GRCh38
Transcripts
NM_181715.2 XM_005244946.1 XM_005244949.2 XM_017000575.1 XM_017000576.1 XM_017000577.1 XM_017000578.1 XM_017000579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3474553 c.1671G>A p.G557G Chr.1: 153949118-153949118 on Build GRCh38
5025362 c.1669G>A p.G557R Chr.1: 153949120-153949120 on Build GRCh38
6507797 c.1569G>T p.Q523H Chr.1: 153949220-153949220 on Build GRCh38
6635778 c.1641C>T p.Y547Y Chr.1: 153949148-153949148 on Build GRCh38
4629747 c.1601C>T p.P534L Chr.1: 153949188-153949188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111395902 Chr.1: 153949008-153949008 on Build GRCh38
rs1000889511 Chr.1: 153949157-153949157 on Build GRCh38
rs770426928 Chr.1: 153949144-153949144 on Build GRCh38
rs1046116901 Chr.1: 153949061-153949061 on Build GRCh38
rs199747428 Chr.1: 153949007-153949007 on Build GRCh38
rs1047288326 Chr.1: 153949047-153949047 on Build GRCh38
rs370354798 Chr.1: 153949183-153949183 on Build GRCh38
rs775864743 Chr.1: 153949075-153949075 on Build GRCh38
rs200292489 Chr.1: 153949137-153949137 on Build GRCh38
rs765463923 Chr.1: 153949121-153949121 on Build GRCh38
rs1014485095 Chr.1: 153949007-153949007 on Build GRCh38
rs1016143816 Chr.1: 153949150-153949150 on Build GRCh38
rs754152872 Chr.1: 153949105-153949105 on Build GRCh38
rs775889536 Chr.1: 153949180-153949180 on Build GRCh38
rs762190511 Chr.1: 153949195-153949195 on Build GRCh38
rs772578093 Chr.1: 153949176-153949176 on Build GRCh38
rs202208580 Chr.1: 153949012-153949012 on Build GRCh38
rs761961350 Chr.1: 153949107-153949107 on Build GRCh38
rs767458793 Chr.1: 153949212-153949212 on Build GRCh38
rs549007441 Chr.1: 153949033-153949033 on Build GRCh38
rs750468582 Chr.1: 153949201-153949201 on Build GRCh38
rs912641868 Chr.1: 153949056-153949056 on Build GRCh38
rs930107846 Chr.1: 153949044-153949044 on Build GRCh38
rs868611934 Chr.1: 153949062-153949062 on Build GRCh38
rs764370887 Chr.1: 153949093-153949093 on Build GRCh38
rs201931769 Chr.1: 153949145-153949145 on Build GRCh38
rs764423895 Chr.1: 153949084-153949085 on Build GRCh38
rs567649028 Chr.1: 153949078-153949078 on Build GRCh38
rs894926012 Chr.1: 153949035-153949035 on Build GRCh38
rs776979010 Chr.1: 153949188-153949188 on Build GRCh38
rs996053568 Chr.1: 153949207-153949207 on Build GRCh38
rs1024170375 Chr.1: 153949037-153949037 on Build GRCh38
rs960862276 Chr.1: 153949003-153949003 on Build GRCh38
rs752630427 Chr.1: 153949216-153949216 on Build GRCh38
rs1054089520 Chr.1: 153949174-153949174 on Build GRCh38
rs762975258 Chr.1: 153949209-153949209 on Build GRCh38
rs113098302 Chr.1: 153949114-153949114 on Build GRCh38
rs200773596 Chr.1: 153949143-153949143 on Build GRCh38
rs143017611 Chr.1: 153949187-153949187 on Build GRCh38
rs528607361 Chr.1: 153949124-153949124 on Build GRCh38
rs963083520 Chr.1: 153949017-153949017 on Build GRCh38
rs781015470 Chr.1: 153949127-153949127 on Build GRCh38
rs765393749 Chr.1: 153949197-153949197 on Build GRCh38
rs777386699 Chr.1: 153949138-153949138 on Build GRCh38
rs745380076 Chr.1: 153949161-153949161 on Build GRCh38
rs752498061 Chr.1: 153949128-153949128 on Build GRCh38
rs974701329 Chr.1: 153949020-153949020 on Build GRCh38
rs754888963 Chr.1: 153949125-153949125 on Build GRCh38

View Full Product Details