Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29321140-29321401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH1
Entrez Gene ID
571
Gene Name
BTB domain and CNC homolog 1
Gene Aliases
BACH-1, BTBD24
UniGene
Hs.154276
Gene Chromosome Location
Chr.21: 29298795-29361896 on Build GRCh38
Transcripts
NM_001186.3 NM_206866.2 NR_027655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357228 c.11G>A p.S4N Chr.21: 29321291-29321291 on Build GRCh38
6609520 c.114C>G p.I38M Chr.21: 29321394-29321394 on Build GRCh38
5792508 c.38C>G p.S13C Chr.21: 29321318-29321318 on Build GRCh38
6609522 c.50G>A p.S17N Chr.21: 29321330-29321330 on Build GRCh38
6609523 c.83G>A p.R28Q Chr.21: 29321363-29321363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910831852 Chr.21: 29321321-29321321 on Build GRCh38
rs373779104 Chr.21: 29321374-29321374 on Build GRCh38
rs374133191 Chr.21: 29321275-29321275 on Build GRCh38
rs771621322 Chr.21: 29321285-29321285 on Build GRCh38
rs1002002126 Chr.21: 29321213-29321213 on Build GRCh38
rs753699279 Chr.21: 29321372-29321372 on Build GRCh38
rs1038132123 Chr.21: 29321344-29321344 on Build GRCh38
rs777733150 Chr.21: 29321261-29321261 on Build GRCh38
rs928011753 Chr.21: 29321193-29321193 on Build GRCh38
rs777643644 Chr.21: 29321367-29321367 on Build GRCh38
rs147482272 Chr.21: 29321358-29321358 on Build GRCh38
rs749193862 Chr.21: 29321270-29321270 on Build GRCh38
rs781710744 Chr.21: 29321297-29321297 on Build GRCh38
rs991123214 Chr.21: 29321209-29321209 on Build GRCh38
rs762065180 Chr.21: 29321300-29321300 on Build GRCh38
rs751579550 Chr.21: 29321329-29321329 on Build GRCh38
rs1028243602 Chr.21: 29321346-29321346 on Build GRCh38
rs141543969 Chr.21: 29321291-29321291 on Build GRCh38
rs747863102 Chr.21: 29321258-29321258 on Build GRCh38
rs756432215 Chr.21: 29321363-29321363 on Build GRCh38
rs368391255 Chr.21: 29321252-29321252 on Build GRCh38
rs201223760 Chr.21: 29321336-29321336 on Build GRCh38
rs773597979 Chr.21: 29321309-29321309 on Build GRCh38
rs778974427 Chr.21: 29321376-29321376 on Build GRCh38
rs1020612695 Chr.21: 29321287-29321287 on Build GRCh38
rs897944831 Chr.21: 29321170-29321170 on Build GRCh38
rs986315857 Chr.21: 29321315-29321315 on Build GRCh38
rs367633782 Chr.21: 29321301-29321301 on Build GRCh38
rs745439507 Chr.21: 29321276-29321276 on Build GRCh38
rs758653110 Chr.21: 29321212-29321212 on Build GRCh38
rs781536847 Chr.21: 29321250-29321250 on Build GRCh38
rs766887188 Chr.21: 29321316-29321316 on Build GRCh38
rs1024984339 Chr.21: 29321206-29321206 on Build GRCh38
rs993560247 Chr.21: 29321195-29321195 on Build GRCh38
rs752987492 Chr.21: 29321207-29321207 on Build GRCh38
rs763385315 Chr.21: 29321311-29321311 on Build GRCh38
rs745600599 Chr.21: 29321258-29321258 on Build GRCh38
rs747087328 Chr.21: 29321362-29321362 on Build GRCh38
rs755725197 Chr.21: 29321275-29321275 on Build GRCh38
rs770992854 Chr.21: 29321273-29321273 on Build GRCh38
rs116727521 Chr.21: 29321260-29321260 on Build GRCh38

View Full Product Details