Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130379934-130380159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A5
Entrez Gene ID
256076
Gene Name
collagen type VI alpha 5
Gene Aliases
COL29A1, VWA4
UniGene
Hs.205403
Gene Chromosome Location
Chr.3: 130345516-130484846 on Build GRCh38
Transcripts
NM_001278298.1 NM_153264.6 NR_022012.2 XM_011512621.2 XM_011512622.2 XM_011512623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6305877 c.1210A>T p.T404S Chr.3: 130379960-130379960 on Build GRCh38
7362301 c.1220C>A p.T407K Chr.3: 130379970-130379970 on Build GRCh38
4461940 c.1227C>T p.F409F Chr.3: 130379977-130379977 on Build GRCh38
5682516 c.1272T>C p.Y424Y Chr.3: 130380022-130380022 on Build GRCh38
6544807 c.1206A>T p.L402F Chr.3: 130379956-130379956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015654380 Chr.3: 130380071-130380071 on Build GRCh38
rs114174330 Chr.3: 130379966-130379966 on Build GRCh38
rs1004623862 Chr.3: 130380061-130380061 on Build GRCh38
rs544127793 Chr.3: 130380082-130380082 on Build GRCh38
rs534863881 Chr.3: 130380085-130380085 on Build GRCh38
rs947065299 Chr.3: 130380089-130380089 on Build GRCh38
rs769844856 Chr.3: 130380112-130380112 on Build GRCh38
rs1042715801 Chr.3: 130380091-130380091 on Build GRCh38
rs902819165 Chr.3: 130380109-130380109 on Build GRCh38
rs1048375006 Chr.3: 130379982-130379982 on Build GRCh38
rs888420716 Chr.3: 130380036-130380036 on Build GRCh38
rs894395053 Chr.3: 130380005-130380005 on Build GRCh38
rs576520883 Chr.3: 130380081-130380081 on Build GRCh38
rs1046246447 Chr.3: 130379965-130379965 on Build GRCh38
rs558238345 Chr.3: 130379975-130379975 on Build GRCh38

View Full Product Details