Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244552478-244552735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf101
Entrez Gene ID
257044
Gene Name
chromosome 1 open reading frame 101
Gene Aliases
-
UniGene
Hs.459534
Gene Chromosome Location
Chr.1: 244452014-244640362 on Build GRCh38
Transcripts
NM_001130957.1 NM_001242340.1 NM_173807.4 XM_011544138.2 XM_011544139.2 XM_011544142.2 XM_011544144.2 XM_011544145.2 XM_011544149.2 XM_011544151.2 XM_017000943.1 XM_017000944.1 XM_017000945.1 XM_017000946.1 XM_017000947.1 XM_017000948.1 XM_017000949.1 XM_017000950.1 XM_017000951.1 XM_017000952.1 XM_017000953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2051335 c.745G>C p.A249P Chr.1: 244552530-244552530 on Build GRCh38
7094682 c.800A>G p.K267R Chr.1: 244552585-244552585 on Build GRCh38
4235258 c.943A>G p.R315G Chr.1: 244552728-244552728 on Build GRCh38
6898219 c.829G>A p.D277N Chr.1: 244552614-244552614 on Build GRCh38
6997869 c.739G>A p.V247I Chr.1: 244552524-244552524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907908869 Chr.1: 244552615-244552615 on Build GRCh38
rs201843159 Chr.1: 244552612-244552612 on Build GRCh38
rs756466860 Chr.1: 244552686-244552686 on Build GRCh38
rs771585112 Chr.1: 244552535-244552535 on Build GRCh38
rs773687728 Chr.1: 244552572-244552572 on Build GRCh38
rs879157986 Chr.1: 244552638-244552638 on Build GRCh38
rs1044413217 Chr.1: 244552663-244552663 on Build GRCh38
rs34510134 Chr.1: 244552580-244552580 on Build GRCh38
rs367552408 Chr.1: 244552662-244552662 on Build GRCh38
rs747847138 Chr.1: 244552656-244552656 on Build GRCh38
rs760368231 Chr.1: 244552545-244552545 on Build GRCh38
rs899435702 Chr.1: 244552636-244552636 on Build GRCh38
rs766477637 Chr.1: 244552579-244552579 on Build GRCh38
rs537004249 Chr.1: 244552616-244552616 on Build GRCh38
rs1052830028 Chr.1: 244552668-244552668 on Build GRCh38
rs758331404 Chr.1: 244552617-244552617 on Build GRCh38
rs138657489 Chr.1: 244552655-244552655 on Build GRCh38
rs113653443 Chr.1: 244552661-244552661 on Build GRCh38
rs147127314 Chr.1: 244552687-244552687 on Build GRCh38
rs557339421 Chr.1: 244552629-244552629 on Build GRCh38
rs749112622 Chr.1: 244552503-244552503 on Build GRCh38
rs767742056 Chr.1: 244552677-244552677 on Build GRCh38
rs538919602 Chr.1: 244552553-244552553 on Build GRCh38
rs768305308 Chr.1: 244552651-244552651 on Build GRCh38
rs755173996 Chr.1: 244552588-244552588 on Build GRCh38
rs767659349 Chr.1: 244552591-244552591 on Build GRCh38
rs752948952 Chr.1: 244552683-244552683 on Build GRCh38
rs776922670 Chr.1: 244552555-244552555 on Build GRCh38
rs759561837 Chr.1: 244552676-244552676 on Build GRCh38
rs779030034 Chr.1: 244552508-244552508 on Build GRCh38
rs746513331 Chr.1: 244552639-244552639 on Build GRCh38
rs770945995 Chr.1: 244552506-244552506 on Build GRCh38
rs377258316 Chr.1: 244552571-244552571 on Build GRCh38
rs567394296 Chr.1: 244552652-244552652 on Build GRCh38
rs371932318 Chr.1: 244552665-244552665 on Build GRCh38
rs35587664 Chr.1: 244552664-244552664 on Build GRCh38
rs745625090 Chr.1: 244552510-244552510 on Build GRCh38
rs774437688 Chr.1: 244552666-244552666 on Build GRCh38
rs566703602 Chr.1: 244552532-244552532 on Build GRCh38
rs763429416 Chr.1: 244552575-244552575 on Build GRCh38
rs753650352 Chr.1: 244552595-244552595 on Build GRCh38
rs774918244 Chr.1: 244552539-244552539 on Build GRCh38

View Full Product Details