Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 85961282-85961513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADM2
Entrez Gene ID
253559
Gene Name
cell adhesion molecule 2
Gene Aliases
IGSF4D, NECL3, Necl-3, SynCAM 2, synCAM2
UniGene
Hs.164578
Gene Chromosome Location
Chr.3: 84958982-86074429 on Build GRCh38
Transcripts
NM_001167674.1 NM_001167675.1 NM_001256502.1 NM_001256503.1 NM_001256504.1 NM_001256505.1 NM_153184.3 XM_006713081.3 XM_006713082.3 XM_017006062.1 XM_017006063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309827 c.794A>T p.K265M Chr.3: 85961492-85961492 on Build GRCh38
1247220 c.823G>A p.G275R Chr.3: 85961500-85961500 on Build GRCh38
331386 c.802G>C p.V268L Chr.3: 85961479-85961479 on Build GRCh38
2827939 c.828A>T p.E276D Chr.3: 85961505-85961505 on Build GRCh38
1247219 c.802G>A p.G268R Chr.3: 85961500-85961500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896589216 Chr.3: 85961373-85961373 on Build GRCh38
rs892593299 Chr.3: 85961379-85961379 on Build GRCh38
rs147616825 Chr.3: 85961479-85961479 on Build GRCh38
rs762651968 Chr.3: 85961477-85961477 on Build GRCh38
rs780397687 Chr.3: 85961429-85961429 on Build GRCh38
rs999990489 Chr.3: 85961358-85961358 on Build GRCh38
rs180853499 Chr.3: 85961337-85961337 on Build GRCh38
rs778875950 Chr.3: 85961442-85961442 on Build GRCh38
rs9819556 Chr.3: 85961450-85961450 on Build GRCh38
rs770807742 Chr.3: 85961449-85961449 on Build GRCh38
rs1002426765 Chr.3: 85961484-85961484 on Build GRCh38
rs902534612 Chr.3: 85961348-85961348 on Build GRCh38
rs1035098229 Chr.3: 85961371-85961371 on Build GRCh38
rs374308520 Chr.3: 85961465-85961465 on Build GRCh38
rs552129698 Chr.3: 85961428-85961428 on Build GRCh38
rs533687349 Chr.3: 85961363-85961363 on Build GRCh38
rs920725898 Chr.3: 85961322-85961322 on Build GRCh38
rs749558125 Chr.3: 85961431-85961431 on Build GRCh38
rs267599940 Chr.3: 85961488-85961488 on Build GRCh38
rs544751415 Chr.3: 85961328-85961336 on Build GRCh38
rs750846935 Chr.3: 85961427-85961427 on Build GRCh38
rs768771561 Chr.3: 85961433-85961433 on Build GRCh38
rs946649471 Chr.3: 85961330-85961330 on Build GRCh38
rs1050677243 Chr.3: 85961329-85961329 on Build GRCh38
rs780060198 Chr.3: 85961446-85961447 on Build GRCh38
rs746900632 Chr.3: 85961445-85961445 on Build GRCh38
rs1012416368 Chr.3: 85961400-85961400 on Build GRCh38
rs775119535 Chr.3: 85961473-85961473 on Build GRCh38

View Full Product Details