Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172570554-172570815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCO
Entrez Gene ID
51430
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1orf9, CH1, OPT, SLP1
UniGene
Hs.204559
Gene Chromosome Location
Chr.1: 172532349-172611833 on Build GRCh38
Transcripts
NM_001282750.1 NM_001282751.1 NM_014283.4 NM_016227.3 XM_006711374.2 XM_006711375.2 XM_006711376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5025432 c.1000A>T p.I334L Chr.1: 172570681-172570681 on Build GRCh38
6122715 c.1459A>T p.S487C Chr.1: 172570663-172570663 on Build GRCh38
899656 c.1037G>A p.S346N Chr.1: 172570718-172570718 on Build GRCh38
5025433 c.1477A>T p.I493L Chr.1: 172570681-172570681 on Build GRCh38
4025121 c.1464A>G p.T488T Chr.1: 172570668-172570668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759199417 Chr.1: 172570749-172570749 on Build GRCh38
rs955550237 Chr.1: 172570638-172570638 on Build GRCh38
rs769940452 Chr.1: 172570659-172570659 on Build GRCh38
rs371841126 Chr.1: 172570763-172570763 on Build GRCh38
rs745973299 Chr.1: 172570774-172570774 on Build GRCh38
rs763469414 Chr.1: 172570682-172570682 on Build GRCh38
rs762427178 Chr.1: 172570728-172570728 on Build GRCh38
rs1025466427 Chr.1: 172570611-172570611 on Build GRCh38
rs763224564 Chr.1: 172570594-172570594 on Build GRCh38
rs745570826 Chr.1: 172570615-172570619 on Build GRCh38
rs771611289 Chr.1: 172570625-172570626 on Build GRCh38
rs746053830 Chr.1: 172570613-172570613 on Build GRCh38
rs775134455 Chr.1: 172570628-172570631 on Build GRCh38
rs765941357 Chr.1: 172570740-172570740 on Build GRCh38
rs757206150 Chr.1: 172570768-172570768 on Build GRCh38
rs764843354 Chr.1: 172570759-172570759 on Build GRCh38
rs886976295 Chr.1: 172570648-172570648 on Build GRCh38
rs921390434 Chr.1: 172570627-172570627 on Build GRCh38
rs751438995 Chr.1: 172570767-172570767 on Build GRCh38
rs769283523 Chr.1: 172570711-172570711 on Build GRCh38
rs1008534098 Chr.1: 172570731-172570731 on Build GRCh38
rs753323639 Chr.1: 172570745-172570745 on Build GRCh38
rs775903232 Chr.1: 172570671-172570671 on Build GRCh38
rs746444017 Chr.1: 172570651-172570652 on Build GRCh38
rs987403921 Chr.1: 172570778-172570778 on Build GRCh38
rs774933127 Chr.1: 172570716-172570716 on Build GRCh38
rs552360801 Chr.1: 172570772-172570772 on Build GRCh38
rs376679357 Chr.1: 172570765-172570765 on Build GRCh38

View Full Product Details