Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36098493-36098758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL8A2
Entrez Gene ID
1296
Gene Name
collagen type VIII alpha 2
Gene Aliases
FECD, FECD1, PPCD, PPCD2
UniGene
Hs.353001
Gene Chromosome Location
Chr.1: 36095236-36126207 on Build GRCh38
Transcripts
NM_001294347.1 NM_005202.3 XM_005270477.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4986097 c.1115G>A p.R372H Chr.1: 36098566-36098566 on Build GRCh38
5719237 c.1030G>A p.E344K Chr.1: 36098651-36098651 on Build GRCh38
4520182 c.1047G>A p.G349G Chr.1: 36098634-36098634 on Build GRCh38
4522213 c.1150G>T p.G384* Chr.1: 36098531-36098531 on Build GRCh38
4891467 c.1037G>T p.G346V Chr.1: 36098644-36098644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765995969 Chr.1: 36098528-36098528 on Build GRCh38
rs778152352 Chr.1: 36098597-36098597 on Build GRCh38
rs879183130 Chr.1: 36098648-36098648 on Build GRCh38
rs756149349 Chr.1: 36098715-36098715 on Build GRCh38
rs543271039 Chr.1: 36098562-36098562 on Build GRCh38
rs987732733 Chr.1: 36098652-36098652 on Build GRCh38
rs780318859 Chr.1: 36098567-36098567 on Build GRCh38
rs763962496 Chr.1: 36098556-36098556 on Build GRCh38
rs774061260 Chr.1: 36098530-36098530 on Build GRCh38
rs374692308 Chr.1: 36098574-36098574 on Build GRCh38
rs376000034 Chr.1: 36098741-36098741 on Build GRCh38
rs770282870 Chr.1: 36098596-36098596 on Build GRCh38
rs771745243 Chr.1: 36098625-36098625 on Build GRCh38
rs778332893 Chr.1: 36098720-36098720 on Build GRCh38
rs752603000 Chr.1: 36098550-36098550 on Build GRCh38
rs752761856 Chr.1: 36098713-36098713 on Build GRCh38
rs370165687 Chr.1: 36098658-36098658 on Build GRCh38
rs536436969 Chr.1: 36098521-36098521 on Build GRCh38
rs759229205 Chr.1: 36098622-36098622 on Build GRCh38
rs757009036 Chr.1: 36098566-36098566 on Build GRCh38
rs372728270 Chr.1: 36098684-36098684 on Build GRCh38
rs930478926 Chr.1: 36098651-36098651 on Build GRCh38
rs781458569 Chr.1: 36098587-36098587 on Build GRCh38
rs768510125 Chr.1: 36098556-36098556 on Build GRCh38
rs748267443 Chr.1: 36098594-36098594 on Build GRCh38
rs781583202 Chr.1: 36098712-36098712 on Build GRCh38
rs749811184 Chr.1: 36098726-36098726 on Build GRCh38
rs760117910 Chr.1: 36098643-36098643 on Build GRCh38
rs773583216 Chr.1: 36098524-36098524 on Build GRCh38
rs774933715 Chr.1: 36098632-36098632 on Build GRCh38
rs764026885 Chr.1: 36098667-36098667 on Build GRCh38
rs755208300 Chr.1: 36098706-36098706 on Build GRCh38
rs199786966 Chr.1: 36098612-36098612 on Build GRCh38
rs749662705 Chr.1: 36098605-36098605 on Build GRCh38
rs1036279189 Chr.1: 36098606-36098606 on Build GRCh38
rs200344795 Chr.1: 36098688-36098688 on Build GRCh38
rs910373367 Chr.1: 36098660-36098660 on Build GRCh38
rs773973727 Chr.1: 36098620-36098620 on Build GRCh38
rs760623325 Chr.1: 36098553-36098553 on Build GRCh38
rs763244654 Chr.1: 36098525-36098525 on Build GRCh38
rs79833067 Chr.1: 36098676-36098676 on Build GRCh38
rs755085660 Chr.1: 36098580-36098580 on Build GRCh38
rs375174124 Chr.1: 36098541-36098541 on Build GRCh38
rs77385288 Chr.1: 36098544-36098544 on Build GRCh38

View Full Product Details