Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63284753-63285018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGAP1
Entrez Gene ID
55738
Gene Name
ADP ribosylation factor GTPase activating protein 1
Gene Aliases
ARF1GAP, HRIHFB2281
UniGene
Hs.25584
Gene Chromosome Location
Chr.20: 63272785-63289793 on Build GRCh38
Transcripts
NM_001281482.1 NM_001281483.1 NM_001281484.1 NM_018209.3 NM_175609.2 NR_104022.1 NR_104023.1 XM_006723823.2 XM_011528901.1 XM_017027950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6890861 c.799G>T p.E267* Chr.20: 63284917-63284917 on Build GRCh38
2931560 c.764C>T p.A255V Chr.20: 63284912-63284912 on Build GRCh38
6602863 c.804+2T>C p.? Chr.20: 63284924-63284924 on Build GRCh38
6276328 c.718-1G>T p.? Chr.20: 63284865-63284865 on Build GRCh38
3963771 c.770A>T p.E257V Chr.20: 63284918-63284918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756423631 Chr.20: 63284968-63284968 on Build GRCh38
rs961735000 Chr.20: 63284857-63284857 on Build GRCh38
rs748820642 Chr.20: 63284919-63284919 on Build GRCh38
rs373944258 Chr.20: 63284863-63284863 on Build GRCh38
rs770390170 Chr.20: 63284922-63284922 on Build GRCh38
rs762633961 Chr.20: 63284874-63284874 on Build GRCh38
rs981320696 Chr.20: 63284988-63284988 on Build GRCh38
rs753784304 Chr.20: 63284937-63284937 on Build GRCh38
rs368033888 Chr.20: 63284867-63284867 on Build GRCh38
rs765445090 Chr.20: 63284891-63284891 on Build GRCh38
rs369800568 Chr.20: 63284882-63284882 on Build GRCh38
rs539741277 Chr.20: 63284963-63284963 on Build GRCh38
rs561650999 Chr.20: 63284810-63284810 on Build GRCh38
rs906936574 Chr.20: 63284772-63284772 on Build GRCh38
rs139966116 Chr.20: 63284916-63284916 on Build GRCh38
rs745817073 Chr.20: 63284961-63284965 on Build GRCh38
rs897246738 Chr.20: 63284979-63284979 on Build GRCh38
rs758659039 Chr.20: 63284851-63284851 on Build GRCh38
rs201358885 Chr.20: 63284824-63284824 on Build GRCh38
rs971288495 Chr.20: 63284985-63284986 on Build GRCh38
rs552463853 Chr.20: 63284847-63284847 on Build GRCh38
rs752634490 Chr.20: 63284887-63284887 on Build GRCh38
rs374739007 Chr.20: 63284974-63284974 on Build GRCh38
rs893059267 Chr.20: 63284796-63284796 on Build GRCh38
rs113430203 Chr.20: 63284809-63284809 on Build GRCh38
rs1053177080 Chr.20: 63284784-63284784 on Build GRCh38
rs766426417 Chr.20: 63284939-63284939 on Build GRCh38
rs372591307 Chr.20: 63284835-63284835 on Build GRCh38
rs200920656 Chr.20: 63284856-63284856 on Build GRCh38
rs771829639 Chr.20: 63284929-63284929 on Build GRCh38
rs757295467 Chr.20: 63284836-63284836 on Build GRCh38
rs149138919 Chr.20: 63284994-63284994 on Build GRCh38
rs147317838 Chr.20: 63284899-63284899 on Build GRCh38
rs112403271 Chr.20: 63284868-63284868 on Build GRCh38
rs994621028 Chr.20: 63284989-63284989 on Build GRCh38
rs79628043 Chr.20: 63284994-63284994 on Build GRCh38
rs781612232 Chr.20: 63284911-63284911 on Build GRCh38
rs368304656 Chr.20: 63284817-63284817 on Build GRCh38
rs372688461 Chr.20: 63284889-63284889 on Build GRCh38
rs748673929 Chr.20: 63284912-63284912 on Build GRCh38
rs140253128 Chr.20: 63284881-63284881 on Build GRCh38
rs759211690 Chr.20: 63284928-63284928 on Build GRCh38
rs376634869 Chr.20: 63284872-63284872 on Build GRCh38
rs750495509 Chr.20: 63284818-63284818 on Build GRCh38
rs201496824 Chr.20: 63284901-63284901 on Build GRCh38
rs370261026 Chr.20: 63284973-63284973 on Build GRCh38
rs751790115 Chr.20: 63284943-63284943 on Build GRCh38
rs73153583 Chr.20: 63284934-63284934 on Build GRCh38
rs781052102 Chr.20: 63284915-63284917 on Build GRCh38
rs375396089 Chr.20: 63284957-63284957 on Build GRCh38
rs369589226 Chr.20: 63284893-63284893 on Build GRCh38
rs770702098 Chr.20: 63284837-63284838 on Build GRCh38
rs767282895 Chr.20: 63284885-63284885 on Build GRCh38
rs746804287 Chr.20: 63284836-63284838 on Build GRCh38
rs780612269 Chr.20: 63284854-63284854 on Build GRCh38
rs767379396 Chr.20: 63284833-63284833 on Build GRCh38
rs374191388 Chr.20: 63284815-63284815 on Build GRCh38
rs62208784 Chr.20: 63284940-63284940 on Build GRCh38
rs202096342 Chr.20: 63284918-63284918 on Build GRCh38
rs934368744 Chr.20: 63284775-63284775 on Build GRCh38
rs375736703 Chr.20: 63284970-63284970 on Build GRCh38
rs755314276 Chr.20: 63284905-63284905 on Build GRCh38
rs528934937 Chr.20: 63284814-63284814 on Build GRCh38
rs765995284 Chr.20: 63284878-63284878 on Build GRCh38
rs924412340 Chr.20: 63284831-63284831 on Build GRCh38
rs143810518 Chr.20: 63284892-63284892 on Build GRCh38
rs759323707 Chr.20: 63284938-63284938 on Build GRCh38
rs770255541 Chr.20: 63284913-63284913 on Build GRCh38
rs1027068845 Chr.20: 63284997-63284997 on Build GRCh38
rs79169534 Chr.20: 63284927-63284927 on Build GRCh38
rs200186934 Chr.20: 63284871-63284871 on Build GRCh38
rs372448777 Chr.20: 63284936-63284936 on Build GRCh38
rs777011259 Chr.20: 63284865-63284865 on Build GRCh38
rs755227399 Chr.20: 63284953-63284953 on Build GRCh38
rs140673126 Chr.20: 63284776-63284776 on Build GRCh38
rs754994305 Chr.20: 63284829-63284829 on Build GRCh38
rs780451093 Chr.20: 63284898-63284898 on Build GRCh38
rs531244036 Chr.20: 63284846-63284846 on Build GRCh38
rs766230506 Chr.20: 63284822-63284822 on Build GRCh38

View Full Product Details