Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12973494-12973719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAND5
Entrez Gene ID
199699
Gene Name
DAN domain BMP antagonist family member 5
Gene Aliases
CER2, CERL2, CKTSF1B3, COCO, CRL2, DANTE, GREM3, SP1
UniGene
Hs.331981
Gene Chromosome Location
Chr.19: 12969618-12974753 on Build GRCh38
Transcripts
NM_152654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5557700 c.458A>C p.K153T Chr.19: 12973522-12973522 on Build GRCh38
6639986 c.509G>A p.R170H Chr.19: 12973573-12973573 on Build GRCh38
5909704 c.454C>T p.R152C Chr.19: 12973518-12973518 on Build GRCh38
3891590 c.507C>T p.S169S Chr.19: 12973571-12973571 on Build GRCh38
6195617 c.472G>A p.V158M Chr.19: 12973536-12973536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs45499495 Chr.19: 12973693-12973693 on Build GRCh38
rs751930538 Chr.19: 12973535-12973535 on Build GRCh38
rs45513495 Chr.19: 12973519-12973519 on Build GRCh38
rs957694460 Chr.19: 12973622-12973622 on Build GRCh38
rs914794424 Chr.19: 12973544-12973544 on Build GRCh38
rs774578194 Chr.19: 12973670-12973670 on Build GRCh38
rs530594712 Chr.19: 12973585-12973585 on Build GRCh38
rs775699908 Chr.19: 12973579-12973579 on Build GRCh38
rs745805814 Chr.19: 12973576-12973576 on Build GRCh38
rs748503801 Chr.19: 12973637-12973640 on Build GRCh38
rs764281812 Chr.19: 12973533-12973533 on Build GRCh38
rs541734703 Chr.19: 12973536-12973536 on Build GRCh38
rs374830367 Chr.19: 12973644-12973644 on Build GRCh38
rs199774491 Chr.19: 12973575-12973575 on Build GRCh38
rs867025906 Chr.19: 12973518-12973518 on Build GRCh38
rs371222553 Chr.19: 12973578-12973578 on Build GRCh38
rs777340186 Chr.19: 12973563-12973563 on Build GRCh38
rs923669240 Chr.19: 12973668-12973668 on Build GRCh38
rs904942902 Chr.19: 12973609-12973609 on Build GRCh38
rs759648370 Chr.19: 12973524-12973524 on Build GRCh38
rs746834132 Chr.19: 12973514-12973514 on Build GRCh38
rs963562114 Chr.19: 12973614-12973614 on Build GRCh38
rs758029495 Chr.19: 12973561-12973561 on Build GRCh38
rs774472060 Chr.19: 12973590-12973590 on Build GRCh38
rs367706500 Chr.19: 12973611-12973611 on Build GRCh38
rs762192599 Chr.19: 12973594-12973594 on Build GRCh38
rs371031823 Chr.19: 12973525-12973525 on Build GRCh38
rs761162804 Chr.19: 12973607-12973607 on Build GRCh38
rs752177513 Chr.19: 12973621-12973621 on Build GRCh38
rs778396935 Chr.19: 12973550-12973550 on Build GRCh38
rs370085230 Chr.19: 12973642-12973642 on Build GRCh38
rs543804820 Chr.19: 12973596-12973596 on Build GRCh38
rs565227010 Chr.19: 12973657-12973657 on Build GRCh38
rs745717544 Chr.19: 12973654-12973654 on Build GRCh38
rs779286827 Chr.19: 12973534-12973535 on Build GRCh38
rs780181111 Chr.19: 12973658-12973658 on Build GRCh38
rs373817477 Chr.19: 12973605-12973605 on Build GRCh38
rs1032427092 Chr.19: 12973619-12973619 on Build GRCh38
rs947633875 Chr.19: 12973562-12973562 on Build GRCh38
rs866965195 Chr.19: 12973617-12973617 on Build GRCh38
rs768645071 Chr.19: 12973666-12973666 on Build GRCh38
rs763265679 Chr.19: 12973528-12973528 on Build GRCh38
rs746786431 Chr.19: 12973572-12973572 on Build GRCh38
rs375577023 Chr.19: 12973573-12973573 on Build GRCh38
rs749429365 Chr.19: 12973584-12973584 on Build GRCh38
rs776471948 Chr.19: 12973520-12973520 on Build GRCh38
rs375137580 Chr.19: 12973659-12973659 on Build GRCh38
rs200839356 Chr.19: 12973604-12973604 on Build GRCh38
rs768031709 Chr.19: 12973537-12973537 on Build GRCh38
rs775790077 Chr.19: 12973526-12973526 on Build GRCh38
rs752267613 Chr.19: 12973553-12973553 on Build GRCh38
rs372018214 Chr.19: 12973646-12973646 on Build GRCh38
rs754429363 Chr.19: 12973539-12973539 on Build GRCh38
rs367951374 Chr.19: 12973645-12973645 on Build GRCh38

View Full Product Details