Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 30709722-30709995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
UniGene
Hs.234355
Gene Chromosome Location
Chr.12: 30709552-30754994 on Build GRCh38
Transcripts
NM_001002259.2 NM_001206856.2 NM_001319842.1 NM_001319843.1 NM_001319844.1 NM_001319845.1 NM_001319846.1 NM_023925.4 NM_032156.4 XM_006719138.3 XM_006719139.3 XM_006719140.3 XM_006719142.3 XM_006719144.3 XM_006719145.3 XM_006719146.3 XM_006719147.3 XM_017019851.1 XM_017019852.1 XM_017019853.1 XM_017019854.1 XM_017019855.1 XM_017019856.1 XM_017019857.1 XM_017019858.1 XM_017019859.1 XM_017019860.1 XM_017019861.1 XM_017019862.1 XM_017019863.1 XM_017019865.1 XM_017019866.1 XM_017019867.1 XM_017019868.1 XM_017019869.1 XM_017019870.1 XM_017019871.1 XM_017019872.1 XM_017019873.1 XM_017019874.1 XM_017019875.1 XM_017019876.1 XM_017019877.1 XM_017019878.1 XM_017019879.1 XM_017019880.1 XM_017019881.1 XM_017019882.1 XM_017019883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3460084 c.3303G>C p.W1101C Chr.12: 30709983-30709983 on Build GRCh38
938645 c.3312G>A p.L1104L Chr.12: 30709974-30709974 on Build GRCh38
938644 c.3354G>A p.T1118T Chr.12: 30709932-30709932 on Build GRCh38
938643 c.3360A>G p.S1120S Chr.12: 30709926-30709926 on Build GRCh38
4895174 c.3370C>T p.L1124F Chr.12: 30709916-30709916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777291280 Chr.12: 30709921-30709921 on Build GRCh38
rs763315430 Chr.12: 30709857-30709857 on Build GRCh38
rs925282811 Chr.12: 30709948-30709948 on Build GRCh38
rs747779716 Chr.12: 30709908-30709908 on Build GRCh38
rs1052436238 Chr.12: 30709969-30709969 on Build GRCh38
rs371420705 Chr.12: 30709953-30709953 on Build GRCh38
rs762896446 Chr.12: 30709806-30709806 on Build GRCh38
rs766398707 Chr.12: 30709933-30709933 on Build GRCh38
rs552563059 Chr.12: 30709932-30709932 on Build GRCh38
rs748225054 Chr.12: 30709773-30709773 on Build GRCh38
rs750299811 Chr.12: 30709865-30709865 on Build GRCh38
rs189968218 Chr.12: 30709873-30709873 on Build GRCh38
rs886569319 Chr.12: 30709938-30709938 on Build GRCh38
rs746415665 Chr.12: 30709956-30709956 on Build GRCh38
rs931902951 Chr.12: 30709860-30709860 on Build GRCh38
rs140573371 Chr.12: 30709804-30709804 on Build GRCh38
rs1043740457 Chr.12: 30709884-30709884 on Build GRCh38
rs563741832 Chr.12: 30709843-30709843 on Build GRCh38
rs771471072 Chr.12: 30709913-30709916 on Build GRCh38
rs767407050 Chr.12: 30709863-30709863 on Build GRCh38
rs555054269 Chr.12: 30709885-30709885 on Build GRCh38
rs899395230 Chr.12: 30709815-30709815 on Build GRCh38
rs770022713 Chr.12: 30709775-30709775 on Build GRCh38
rs754447478 Chr.12: 30709887-30709887 on Build GRCh38
rs779984888 Chr.12: 30709882-30709882 on Build GRCh38
rs778437920 Chr.12: 30709891-30709891 on Build GRCh38

View Full Product Details