Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33016425-33016698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC7
Entrez Gene ID
91614
Gene Name
DEP domain containing 7
Gene Aliases
TR2, dJ85M6.4
UniGene
Hs.280990
Gene Chromosome Location
Chr.11: 33015864-33033582 on Build GRCh38
Transcripts
NM_001077242.1 NM_139160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3787045 c.32A>G p.E11G Chr.11: 33016571-33016571 on Build GRCh38
2114794 c.41T>C p.I14T Chr.11: 33016580-33016580 on Build GRCh38
6353347 c.3G>A p.M1I Chr.11: 33016542-33016542 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752554581 Chr.11: 33016577-33016577 on Build GRCh38
rs776795886 Chr.11: 33016537-33016537 on Build GRCh38
rs761789836 Chr.11: 33016628-33016630 on Build GRCh38
rs1043828071 Chr.11: 33016469-33016469 on Build GRCh38
rs995250260 Chr.11: 33016598-33016598 on Build GRCh38
rs945485173 Chr.11: 33016448-33016448 on Build GRCh38
rs747313449 Chr.11: 33016531-33016531 on Build GRCh38
rs374161301 Chr.11: 33016592-33016592 on Build GRCh38
rs777516918 Chr.11: 33016489-33016489 on Build GRCh38
rs372349701 Chr.11: 33016632-33016632 on Build GRCh38
rs772381311 Chr.11: 33016518-33016518 on Build GRCh38
rs773347974 Chr.11: 33016628-33016628 on Build GRCh38
rs768891531 Chr.11: 33016532-33016532 on Build GRCh38
rs1037628779 Chr.11: 33016587-33016587 on Build GRCh38
rs758955396 Chr.11: 33016610-33016610 on Build GRCh38
rs761212330 Chr.11: 33016483-33016483 on Build GRCh38
rs766755267 Chr.11: 33016552-33016552 on Build GRCh38
rs749323829 Chr.11: 33016525-33016526 on Build GRCh38
rs368000196 Chr.11: 33016629-33016629 on Build GRCh38
rs577244158 Chr.11: 33016579-33016579 on Build GRCh38
rs370129750 Chr.11: 33016660-33016660 on Build GRCh38
rs570187003 Chr.11: 33016458-33016458 on Build GRCh38
rs79223825 Chr.11: 33016622-33016622 on Build GRCh38
rs780493194 Chr.11: 33016614-33016614 on Build GRCh38
rs765587884 Chr.11: 33016667-33016667 on Build GRCh38
rs371138729 Chr.11: 33016505-33016505 on Build GRCh38
rs1001579218 Chr.11: 33016642-33016642 on Build GRCh38
rs769860928 Chr.11: 33016605-33016605 on Build GRCh38
rs925283389 Chr.11: 33016475-33016475 on Build GRCh38
rs1018803696 Chr.11: 33016668-33016668 on Build GRCh38
rs748388900 Chr.11: 33016623-33016623 on Build GRCh38
rs774585517 Chr.11: 33016531-33016532 on Build GRCh38
rs538975516 Chr.11: 33016492-33016492 on Build GRCh38
rs769831778 Chr.11: 33016627-33016627 on Build GRCh38
rs374778072 Chr.11: 33016510-33016510 on Build GRCh38
rs748837645 Chr.11: 33016490-33016490 on Build GRCh38
rs747407686 Chr.11: 33016620-33016620 on Build GRCh38
rs200779852 Chr.11: 33016560-33016560 on Build GRCh38
rs199606368 Chr.11: 33016599-33016599 on Build GRCh38
rs938016559 Chr.11: 33016464-33016464 on Build GRCh38
rs769898752 Chr.11: 33016551-33016551 on Build GRCh38
rs761842467 Chr.11: 33016543-33016543 on Build GRCh38
rs780399175 Chr.11: 33016520-33016520 on Build GRCh38
rs767293522 Chr.11: 33016571-33016571 on Build GRCh38
rs775297931 Chr.11: 33016498-33016498 on Build GRCh38
rs1035350019 Chr.11: 33016652-33016652 on Build GRCh38
rs886375138 Chr.11: 33016558-33016558 on Build GRCh38
rs753518618 Chr.11: 33016586-33016586 on Build GRCh38
rs775838279 Chr.11: 33016502-33016503 on Build GRCh38
rs367845306 Chr.11: 33016526-33016526 on Build GRCh38
rs539823514 Chr.11: 33016585-33016585 on Build GRCh38
rs768712378 Chr.11: 33016530-33016530 on Build GRCh38

View Full Product Details