Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23249171-23249430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP7
Entrez Gene ID
91782
Gene Name
charged multivesicular body protein 7
Gene Aliases
-
UniGene
Hs.5019
Gene Chromosome Location
Chr.8: 23243637-23262000 on Build GRCh38
Transcripts
NM_001317899.1 NM_152272.4 XM_006716415.2 XM_017013961.1 XM_017013962.1 XM_017013963.1 XM_017013964.1 XM_017013965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2786575 c.334G>A p.A112T Chr.8: 23249244-23249244 on Build GRCh38
7280680 c.394A>G p.K132E Chr.8: 23249304-23249304 on Build GRCh38
4672189 c.361_362insG p.V123fs*15 Chr.8: 23249271-23249272 on Build GRCh38
3899009 c.317G>A p.R106Q Chr.8: 23249227-23249227 on Build GRCh38
3720191 c.403C>G p.L135V Chr.8: 23249313-23249313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761532888 Chr.8: 23249391-23249391 on Build GRCh38
rs369022160 Chr.8: 23249388-23249388 on Build GRCh38
rs577891916 Chr.8: 23249397-23249397 on Build GRCh38
rs751479818 Chr.8: 23249340-23249340 on Build GRCh38
rs757899112 Chr.8: 23249323-23249323 on Build GRCh38
rs761321257 Chr.8: 23249305-23249305 on Build GRCh38
rs749814820 Chr.8: 23249235-23249235 on Build GRCh38
rs749433542 Chr.8: 23249356-23249356 on Build GRCh38
rs373965840 Chr.8: 23249289-23249289 on Build GRCh38
rs773105548 Chr.8: 23249231-23249231 on Build GRCh38
rs201155473 Chr.8: 23249410-23249410 on Build GRCh38
rs766361757 Chr.8: 23249335-23249335 on Build GRCh38
rs749871001 Chr.8: 23249317-23249317 on Build GRCh38
rs769951659 Chr.8: 23249227-23249227 on Build GRCh38
rs771109211 Chr.8: 23249357-23249357 on Build GRCh38
rs758275144 Chr.8: 23249203-23249203 on Build GRCh38
rs149327190 Chr.8: 23249347-23249347 on Build GRCh38
rs966430188 Chr.8: 23249204-23249204 on Build GRCh38
rs754846276 Chr.8: 23249341-23249341 on Build GRCh38
rs767741483 Chr.8: 23249296-23249296 on Build GRCh38
rs760026994 Chr.8: 23249292-23249292 on Build GRCh38
rs762499978 Chr.8: 23249399-23249399 on Build GRCh38
rs774982077 Chr.8: 23249288-23249288 on Build GRCh38
rs34347119 Chr.8: 23249400-23249400 on Build GRCh38
rs764819133 Chr.8: 23249313-23249313 on Build GRCh38
rs139229850 Chr.8: 23249212-23249212 on Build GRCh38
rs781530482 Chr.8: 23249224-23249224 on Build GRCh38
rs376891018 Chr.8: 23249363-23249363 on Build GRCh38
rs746336236 Chr.8: 23249373-23249373 on Build GRCh38
rs780813005 Chr.8: 23249344-23249344 on Build GRCh38
rs776375203 Chr.8: 23249301-23249301 on Build GRCh38
rs369469448 Chr.8: 23249199-23249199 on Build GRCh38
rs771628420 Chr.8: 23249255-23249255 on Build GRCh38
rs756220811 Chr.8: 23249348-23249348 on Build GRCh38
rs541361637 Chr.8: 23249354-23249354 on Build GRCh38
rs372480929 Chr.8: 23249389-23249389 on Build GRCh38
rs779830659 Chr.8: 23249205-23249205 on Build GRCh38
rs112158063 Chr.8: 23249219-23249219 on Build GRCh38
rs748289598 Chr.8: 23249226-23249226 on Build GRCh38

View Full Product Details