Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 114628495-114628722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP2
Entrez Gene ID
93986
Gene Name
forkhead box P2
Gene Aliases
CAGH44, SPCH1, TNRC10
UniGene
Hs.282787
Gene Chromosome Location
Chr.7: 114086310-114693772 on Build GRCh38
Transcripts
NM_001172766.2 NM_001172767.2 NM_014491.3 NM_148898.3 NM_148899.3 NM_148900.3 NR_033766.1 NR_033767.1 XM_017012801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3922523 c.112C>T p.L38L Chr.7: 114628669-114628669 on Build GRCh38
85073 c.467A>G p.Q156R Chr.7: 114628673-114628673 on Build GRCh38
4544064 c.273G>A p.V91V Chr.7: 114628554-114628554 on Build GRCh38
6176001 c.357G>A p.M119I Chr.7: 114628563-114628563 on Build GRCh38
7111834 c.271G>A p.V91M Chr.7: 114628552-114628552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868366293 Chr.7: 114628555-114628555 on Build GRCh38
rs74507296 Chr.7: 114628571-114628571 on Build GRCh38
rs931162237 Chr.7: 114628641-114628641 on Build GRCh38
rs1025011360 Chr.7: 114628554-114628554 on Build GRCh38
rs75306477 Chr.7: 114628569-114628569 on Build GRCh38
rs374479957 Chr.7: 114628686-114628686 on Build GRCh38
rs780212480 Chr.7: 114628580-114628580 on Build GRCh38
rs746703454 Chr.7: 114628603-114628603 on Build GRCh38
rs972636438 Chr.7: 114628522-114628522 on Build GRCh38
rs748583345 Chr.7: 114628611-114628611 on Build GRCh38
rs142280302 Chr.7: 114628608-114628608 on Build GRCh38
rs751403410 Chr.7: 114628557-114628557 on Build GRCh38
rs886061915 Chr.7: 114628623-114628623 on Build GRCh38
rs756184095 Chr.7: 114628574-114628574 on Build GRCh38
rs908446199 Chr.7: 114628661-114628661 on Build GRCh38
rs752334218 Chr.7: 114628536-114628536 on Build GRCh38
rs772558064 Chr.7: 114628614-114628614 on Build GRCh38
rs749418942 Chr.7: 114628581-114628581 on Build GRCh38
rs769148228 Chr.7: 114628669-114628669 on Build GRCh38
rs763871589 Chr.7: 114628693-114628693 on Build GRCh38
rs773759127 Chr.7: 114628632-114628632 on Build GRCh38
rs140009033 Chr.7: 114628596-114628596 on Build GRCh38
rs762699659 Chr.7: 114628549-114628549 on Build GRCh38
rs797045587 Chr.7: 114628563-114628563 on Build GRCh38
rs757255710 Chr.7: 114628564-114628564 on Build GRCh38
rs80073025 Chr.7: 114628653-114628653 on Build GRCh38
rs750505654 Chr.7: 114628573-114628573 on Build GRCh38
rs763737160 Chr.7: 114628550-114628550 on Build GRCh38
rs762445247 Chr.7: 114628688-114628688 on Build GRCh38

View Full Product Details