Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151924401-151924668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA1
Entrez Gene ID
2741
Gene Name
glycine receptor alpha 1
Gene Aliases
HKPX1, STHE
UniGene
Hs.121490
Gene Chromosome Location
Chr.5: 151820798-151924836 on Build GRCh38
Transcripts
NM_000171.3 NM_001146040.1 NM_001292000.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4487535 c.31C>T p.L11F Chr.5: 151924519-151924519 on Build GRCh38
3919398 c.12C>T p.F4F Chr.5: 151924538-151924538 on Build GRCh38
1064665 c.22C>T p.R8* Chr.5: 151924528-151924528 on Build GRCh38
3696835 c.51C>A p.F17L Chr.5: 151924499-151924499 on Build GRCh38
4910697 c.49T>C p.F17L Chr.5: 151924501-151924501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760969146 Chr.5: 151924505-151924505 on Build GRCh38
rs752436268 Chr.5: 151924475-151924475 on Build GRCh38
rs781124134 Chr.5: 151924474-151924474 on Build GRCh38
rs200604203 Chr.5: 151924532-151924532 on Build GRCh38
rs377246222 Chr.5: 151924453-151924453 on Build GRCh38
rs769149313 Chr.5: 151924575-151924575 on Build GRCh38
rs745402231 Chr.5: 151924548-151924548 on Build GRCh38
rs754926052 Chr.5: 151924467-151924467 on Build GRCh38
rs535909506 Chr.5: 151924442-151924442 on Build GRCh38
rs930816700 Chr.5: 151924615-151924615 on Build GRCh38
rs770005385 Chr.5: 151924590-151924590 on Build GRCh38
rs761034478 Chr.5: 151924446-151924446 on Build GRCh38
rs775732411 Chr.5: 151924444-151924444 on Build GRCh38
rs376426309 Chr.5: 151924500-151924500 on Build GRCh38
rs867092651 Chr.5: 151924648-151924648 on Build GRCh38
rs985575972 Chr.5: 151924449-151924449 on Build GRCh38
rs940800574 Chr.5: 151924567-151924567 on Build GRCh38
rs762105396 Chr.5: 151924589-151924589 on Build GRCh38
rs74542605 Chr.5: 151924527-151924527 on Build GRCh38
rs202202750 Chr.5: 151924560-151924560 on Build GRCh38
rs776026097 Chr.5: 151924503-151924503 on Build GRCh38
rs965015586 Chr.5: 151924454-151924454 on Build GRCh38
rs769117123 Chr.5: 151924506-151924506 on Build GRCh38
rs889585221 Chr.5: 151924640-151924640 on Build GRCh38
rs141704405 Chr.5: 151924511-151924511 on Build GRCh38
rs755895077 Chr.5: 151924478-151924478 on Build GRCh38
rs773379170 Chr.5: 151924592-151924592 on Build GRCh38
rs748997223 Chr.5: 151924490-151924490 on Build GRCh38
rs886060281 Chr.5: 151924629-151924629 on Build GRCh38
rs1007219983 Chr.5: 151924422-151924422 on Build GRCh38
rs754014450 Chr.5: 151924458-151924458 on Build GRCh38
rs777041288 Chr.5: 151924584-151924584 on Build GRCh38
rs373393975 Chr.5: 151924489-151924489 on Build GRCh38
rs770675720 Chr.5: 151924491-151924491 on Build GRCh38
rs188720348 Chr.5: 151924441-151924441 on Build GRCh38
rs1036631126 Chr.5: 151924602-151924602 on Build GRCh38
rs777749834 Chr.5: 151924538-151924538 on Build GRCh38
rs747595543 Chr.5: 151924563-151924563 on Build GRCh38
rs753680915 Chr.5: 151924540-151924540 on Build GRCh38
rs1026908572 Chr.5: 151924631-151924631 on Build GRCh38
rs978805775 Chr.5: 151924455-151924455 on Build GRCh38
rs779413449 Chr.5: 151924559-151924559 on Build GRCh38
rs778631570 Chr.5: 151924492-151924492 on Build GRCh38
rs750512870 Chr.5: 151924519-151924519 on Build GRCh38
rs377268097 Chr.5: 151924556-151924556 on Build GRCh38
rs762004647 Chr.5: 151924459-151924459 on Build GRCh38
rs543726938 Chr.5: 151924643-151924643 on Build GRCh38
rs373114110 Chr.5: 151924535-151924535 on Build GRCh38
rs199989616 Chr.5: 151924466-151924466 on Build GRCh38
rs368269982 Chr.5: 151924604-151924604 on Build GRCh38
rs765344212 Chr.5: 151924460-151924460 on Build GRCh38
rs745317657 Chr.5: 151924498-151924498 on Build GRCh38
rs764255064 Chr.5: 151924451-151924451 on Build GRCh38
rs995824748 Chr.5: 151924625-151924625 on Build GRCh38
rs1018856518 Chr.5: 151924427-151924427 on Build GRCh38
rs757026949 Chr.5: 151924545-151924545 on Build GRCh38
rs572343839 Chr.5: 151924508-151924508 on Build GRCh38
rs370920306 Chr.5: 151924525-151924525 on Build GRCh38
rs778643926 Chr.5: 151924546-151924546 on Build GRCh38

View Full Product Details