Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 62747274-62747535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHBP1
Entrez Gene ID
23301
Gene Name
EH domain binding protein 1
Gene Aliases
HPC12, NACSIN
UniGene
Hs.271667
Gene Chromosome Location
Chr.2: 62673851-63046487 on Build GRCh38
Transcripts
NM_001142614.1 NM_001142615.2 NM_001142616.1 NM_015252.3 XM_005264225.2 XM_005264226.2 XM_005264227.1 XM_005264228.2 XM_006711971.2 XM_011532713.2 XM_011532714.2 XM_011532715.2 XM_011532716.2 XM_011532717.1 XM_011532718.2 XM_017003643.1 XM_017003644.1 XM_017003645.1 XM_017003646.1 XM_017003647.1 XM_017003648.1 XM_017003649.1 XM_017003650.1 XM_017003651.1 XM_017003652.1 XM_017003653.1 XM_017003654.1 XM_017003655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752605 c.105-2A>C p.? Chr.2: 62747393-62747393 on Build GRCh38
5397206 c.124G>A p.V42I Chr.2: 62747414-62747414 on Build GRCh38
6652381 c.105G>C p.W35C Chr.2: 62747395-62747395 on Build GRCh38
3839805 c.122T>G p.V41G Chr.2: 62747412-62747412 on Build GRCh38
3054761 c.140G>T p.R47I Chr.2: 62747430-62747430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029320030 Chr.2: 62747425-62747425 on Build GRCh38
rs575950973 Chr.2: 62747328-62747328 on Build GRCh38
rs370536624 Chr.2: 62747437-62747437 on Build GRCh38
rs12465326 Chr.2: 62747391-62747391 on Build GRCh38
rs752356881 Chr.2: 62747418-62747418 on Build GRCh38
rs766413842 Chr.2: 62747365-62747365 on Build GRCh38
rs1007133319 Chr.2: 62747435-62747435 on Build GRCh38
rs767291178 Chr.2: 62747350-62747350 on Build GRCh38
rs751842973 Chr.2: 62747443-62747443 on Build GRCh38
rs1003261539 Chr.2: 62747427-62747427 on Build GRCh38
rs768507702 Chr.2: 62747382-62747386 on Build GRCh38
rs769822005 Chr.2: 62747500-62747500 on Build GRCh38
rs776251241 Chr.2: 62747426-62747428 on Build GRCh38
rs923534626 Chr.2: 62747447-62747447 on Build GRCh38
rs561573225 Chr.2: 62747334-62747334 on Build GRCh38
rs746813164 Chr.2: 62747377-62747378 on Build GRCh38
rs200170310 Chr.2: 62747392-62747392 on Build GRCh38
rs979521049 Chr.2: 62747385-62747385 on Build GRCh38
rs781657751 Chr.2: 62747482-62747482 on Build GRCh38
rs757690206 Chr.2: 62747470-62747470 on Build GRCh38
rs543343236 Chr.2: 62747333-62747333 on Build GRCh38
rs565307490 Chr.2: 62747429-62747429 on Build GRCh38
rs373150728 Chr.2: 62747371-62747371 on Build GRCh38
rs758290512 Chr.2: 62747376-62747377 on Build GRCh38
rs932248000 Chr.2: 62747456-62747456 on Build GRCh38
rs779841785 Chr.2: 62747377-62747377 on Build GRCh38
rs760583185 Chr.2: 62747353-62747353 on Build GRCh38
rs747845187 Chr.2: 62747478-62747478 on Build GRCh38
rs746390309 Chr.2: 62747489-62747489 on Build GRCh38
rs777532284 Chr.2: 62747438-62747438 on Build GRCh38

View Full Product Details